Skip to Content
Merck
All Photos(1)

Key Documents

ABE1953

Sigma-Aldrich

Anti-PTRF/Cavin-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Polymerase I and transcript release factor, PTRF/Cavin-1, Cavin-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$563.00

$563.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$563.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$563.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CAVIN1(284119)

General description

Polymerase I and transcript release factor (UniProt Q6NZI2; also known as Cavin-1, PTRF, RNA polymerase I and transcript release factor, TTF-I interacting peptide 12) is encoded by the PTRF (also known as CGL4, CAVIN, CAVIN1, FKSG13) gene (Gene ID 284119) in human. Cavin-1 is the founding member of the PTRF/Cavin family of cytoplasmic proteins whose expressions are essential for caveola formation. PTRF/Cavin-1, serum deprivation response (SDR)/Cavin-2, SDR-related gene product that binds to C kinase (SRBC)/Cavin-3, and muscle-restricted coiled-coil protein (MURC)/Cavin-4, form an oligomeric assembly termed Cavin complex in the cytosol and associate with caveolin at plasma membrane caveolae. Research shows an essential role for PTRF/Cavin-1 but not the other family members in caveola formation and recruitment of the cavin complex to the plasma membrane. In prostate cancer PC3 cells and during development of zebrafish notochord, lack of PTRF-Cavin expression correlates with lack of caveolae, where caveolin displays increased lateral mobility on the plasma membrane and accelerated lysosomal degradation. Expression of Cavin-1 restores caveolae formation in PC3 cells. Likewise the absence of cavin-1 is also observed in advanced prostate carcinoma. PTRF gene mutations are known to cause congenital generalized lipodystrophy 4 (CGL4).

Specificity

Expected to react with spliced isoforms 1 and 2, but not isoform 3.

Immunogen

Linear peptide corresponding to the N-terminus of human PTRF/Cavin-1.

Application

Anti-PTRF/Cavin-1 Antibody is an antibody against PTRF/Cavin-1 for use in Western Blotting, Immunohistochemistry (Paraffin), Immunocytochemistry.
Western Blotting Analysis: A representative lot detected PTRF/Cavin-1 expression in HeLa, but not in prostate cancer cell lines, LNCaP, 22Rv1 and PC-3 (Moon, H., et al. (2014). Oncogene 33(27):3561-3570).
Western Blotting Analysis: A representative lot detected exogenously expressed human PTRF/Cavin-1 in transfected 22Rv1 and LNCaP cells (Moon, H., et al. (2014). Oncogene 33(27):3561-3570).
Immunohistochemistry Analysis: A representative lot detected reduced PTRF/Cavin-1 immunoreactivity in paraffin-embedded malignant prostate stroma tissue samples when compared with normal prostate stroma and benign prostatic hyperplasia (BPH) specimens. A lack of PTRF/cavin-1 immunoreactivity in prostate epithelia was seen among both normal and malignant samples (Moon, H., et al. (2014). Oncogene 33(27):3561-3570).
Immunocytochemistry Analysis: A representative lot detected surface caveola localization of exogenously expressed human PTRF/Cavin-1 among transfected LNCaP cells (Moon, H., et al. (2014). Oncogene 33(27):3561-3570).

Quality

Evaluated by Western Blotting in rat skeletal muscle myoblast L6 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PTRF/Cavin-1 in 10 µg of rat skeletal muscle myoblast L6 cell lysate.

Target description

~52 kDa observed.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bayader Annabi et al.
International journal of stem cells, 10(1), 103-113 (2016-12-28)
Tumour necrosis factor (TNF)-α activation of mesenchymal stromal cells (MSC) enhances their tumour-suppressive properties and tumour-homing ability. The molecular actors involved are unknown. We found that TNF induced MSC migration and tubulogenesis which correlated with a dose-dependent increase in Cavin-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service