Skip to Content
Merck
All Photos(1)

Key Documents

ABE1950

Sigma-Aldrich

Anti-DBC1 Antibody

from rabbit

Synonym(s):

Cell cycle and apoptosis regulator protein 2, Cell division cycle and apoptosis regulator protein 2, DBIRD complex subunit KIAA1967, Deleted in breast cancer gene 1 protein, DBC-1, DBC.1, p30 DBC, DBC1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$527.00

$527.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$527.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$527.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCAR2(57805)

General description

Cell cycle and apoptosis regulator protein 2 (UniProt Q8N163; also known as Cell division cycle and apoptosis regulator protein 2, DBC-1, DBC.1, DBIRD complex subunit KIAA1967, Deleted in breast cancer 1, p30 DBC, p30 DBC protein) is encoded by the CCAR2 (also known as DBC1, KIAA1967, NET35) gene (Gene ID 57805) in human. Deleted in breast cancer gene 1 protein (DBC1) is a nuclear protein involved in regulating functions of nuclear receptors (NRs), including androgen receptor (AR) and estrogen receptor (ER) alpha and beta. DBC1 stabilizes the interaction between chicken ovalbumin upstream promoter transcription factor I (COUP-TFI) and NCoR by interacting directly with both proteins. DBC1 also interacts with NR complex interacting factor (NRC)-1 to drive the transcription regulation of NRs. Caspase-dependent processing of DBC1 promotes apoptosis, and depletion of DBC1 negatively regulates p53-dependent apoptosis through its specific inhibition of SIRT1. The N-terimus of DBC1 is shown to interact with BRCA1 C-terminal BRCT domain and the expression of DBC1 represses BRCA1-mediated transactivation of the SIRT1 promoter. DBC1 is also a negative regulator against the ligand-dependent transcriptional activation function of ERbeta. The DBC1 protein encoded by the CCAR2 gene is not the same as the protein encoded by the BRINP1 gene, which also goes by the same acronym DBC1 for "Deleted in bladder cancer protein 1".

Specificity

Expected to reacts with both spliced isoforms.

Immunogen

GST-tagged recombinant protein corresponding to human DBC1.

Application

Detect DBC1 using this Anti-DBC1 Antibody validated for use in Western Blotting, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: A representative lot detected DBC1 in HeLa cell lysate (Hiraike, H., et al. (2010). British Journal of Cancer. 102:1061-1067).
Western Blotting Analysis: A representative lot detected DBC1 in MDA-MB-231 cell lysate (Koyama, S., et al. (2010). BBRC. 392:357-362).
Immunoprecipitation Analysis: A representative lot immunoprecipitated DBC1 in HeLa cell lysate (Hiraike, H., et al. (2010). British Journal of Cancer. 102:1061-1067).
Immunocytochemistry Analysis: A representative lot detected DBC1 in MCF7 cells (Hiraike, H., et al. (2010). British Journal of Cancer. 102:1061-1067).
Immunocytochemistry Analysis: A representative lot detected DBC1 in MCF7 and T47D cells (Koyama, S., et al. (2010). BBRC. 392:357-362).
Immunohistochemistry Analysis: A representative lot detected DBC1 in ductal epithelium and adipose regions of human breast tissue sections (Hiraike, H., et al. (2010). British Journal of Cancer. 102:1061-1067).
Immunohistochemistry Analysis: A representative lot detected DBC1 in human breast cancer tissue (Hiraike, H., et al. (2011). Experimental and Therapeutic Medicane. 2:1105-1109).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected DBC1 occupancy at the SIRT1 gene promoter region (1354–1902) known to be targeted by BRCA1 (Hiraike, H., et al. (2010). Br. J. Cancer. 102(6):1061-1067).

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected DBC1 in 10 µg of MCF7 cell lysate.

Target description

~130 kDa observed

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service