ABE1828
Anti-FOXK2 Antibody
serum, from rabbit
Synonym(s):
Forkhead box protein K2, Cellular transcription factor ILF-1, FOXK1, Interleukin enhancer-binding factor 1, FOXK2
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FOXK2(3607)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected FOXK2 in tag-affinity purified BAP1 complex from HeLa cells expressing Flag-HA-tagged wild-type BAP1 or catalytically dead C91S BAP1 mutant (Mashtalir, N., et al. (2014). Mol. Cell. 54(3):392-406).
Epigenetics & Nuclear Function
Nuclear Receptors
Quality
Western Blotting Analysis: A 1:500 dilution of this antibody detected FOXK2 in 10 µg of HeLa nuclear extract.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service