Skip to Content
Merck
All Photos(1)

Key Documents

ABE149

Sigma-Aldrich

Anti-ASF1a Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone chaperone ASF1A, Anti-silencing function protein 1 homolog A, hAsf1, hAsf1a, CCG1-interacting factor A, CIA, hCIA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$743.00

$743.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASF1A(25842)

General description

ASF1a (anti-silencing function 1a) is a histone H3/H4 chaperone and chromatin regulator. This protein facilitates histone placement and histone removal and exchange during nucleosome assembly/disassembly and is essential for the formation of senescence-associated heterochromatic foci (SAHF). ASF1a interacts with chromatin assembly factor 1 (CAF-1) to promote replication-dependent chromatin assembly and with HIRA inside PML bodies of cells approaching senescence after p53/p21 activation.

Immunogen

GST-tagged recombinant protein corresponding to human ASF1a.

Application

Anti-ASF1a Antibody is a Rabbit Polyclonal Antibody for detection of ASF1a also known as Histone chaperone ASF1A or hAsf1 & has been validated in WB & IP.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in IP. (Banumathy, G, et al. (2009). Mol Cell Biol. 29(3):758–770.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected ASF1a on 10 µg of HeLa cell lysate.

Target description

~23 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankit Jaiswal et al.
Scientific reports, 13(1), 19615-19615 (2023-11-11)
Activation-induced cytidine deaminase (AID) is the key mediator of antibody diversification in activated B-cells by the process of somatic hypermutation (SHM) and class switch recombination (CSR). Targeting AID to the Ig genes requires transcription (initiation and elongation), enhancers, and its
Shangda Yang et al.
Nature communications, 9(1), 1285-1285 (2018-03-31)
Histone chaperone ASF1A has been reported to be dysregulated in multiple tumors; however, the underlying molecular mechanism that how the abundance and function of ASF1A are regulated remains unclear. Here we report that ASF1A is physically associated with USP52, which
Chaoyang Xiong et al.
BMC biology, 16(1), 110-110 (2018-10-05)
H3.3 is an ancient and conserved H3 variant and plays essential roles in transcriptional regulation. HIRA complex, which is composed of HIRA, UBN1 or UBN2, and Cabin1, is a H3.3 specific chaperone complex. However, it still remains largely uncharacterized how

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service