Skip to Content
Merck
All Photos(2)

Key Documents

ABE1438

Sigma-Aldrich

Anti-60S Ribosomal Protein L26

from rabbit, purified by affinity chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RPL26(6154)

General description

60S ribosomal protein L26 (UniProt: P61254) is encoded by the RPL26 gene (Gene ID: 6154) in human. 60S ribosomal protein L26 belongs to the ribosomal protein L24P family and is found in all eukaryotes. It is primarily localized in the cytoplasm, but to a lesser extent it is also found in the nucleoplasm and nucleolus. It associates with the large 60S subunit of the ribosome and contains a KOW motif, a conserved RNA-binding motif present in many ribosomal proteins. 60S ribosomal protein L26 is shown to be involved in translation. Knockdown of RPL26 results in a significant decrease in synthesis of 28S rRNA (associated with 60S subunit). Overexpression of RPL26 is shown to enhance the binding of p53 mRNA to the ribosomes and increase p53 translation, which subsequently leads to cell-cycle arrest at G1 phase and can enhance radiation-stimulated apoptosis. Mutations in RPL26 gene are reported to cause congenital non-regenerative hypoplastic anemia that is characterized by a moderate to severe macrocytic anemia, erythroblastopenia, and an increased risk of malignancy.

Specificity

This polyclonal antibody detects 60S ribosomal protein L26 in human.

Immunogen

GST-tagged recombinant fragment corresponding to the first 95 amino acids from human 60S ribosomal protein L26.

Application

Anti-60S Ribosomal Protein L26 Antibody, Cat. No. ABE1438, is a highly specific rabbit polyclonal antibody that targets 60S ribosomal protein L26 and has been tested in Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected 60S Ribosomal Protein L26 in human cerebral cortex, human pancreas, and human colon tissues.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in Hek293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected 60S Ribosomal Protein L26 in 10 µg of HEK293 cell lysate.

Target description

~17 kDa observed; 17.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service