Skip to Content
Merck
All Photos(1)

Key Documents

ABE1424

Sigma-Aldrich

Anti-Glucocorticoid Receptor Antibody

serum, from rabbit

Synonym(s):

Glucocorticoid Receptor, GR, Nuclear receptor subfamily 3 group C member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$479.00

$479.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$479.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$479.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

nonhuman primates (based on 100% sequence homology), canine (based on 100% sequence homology), bovine (based on 100% sequence homology), feline (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

affinity binding assay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NR3C1(2908)

General description

Glucocorticoid receptor (UniProt P04150; also known as GR, Nuclear receptor subfamily 3 group C member 1) is encoded by the NR3C1 (also known as GCCR, GCR, GCRST, GRL) gene (Gene ID 2908) in human. Glucocorticoid receptor is widely expressed in almost all cell types and regulates genes controlling development, metabolism, and immune response upon binding glucocorticoids. Hormone-bound GR translocates from the cytoplasm to the nucleus where it regulates gene transcriptions by binding to glucocorticoid response elements (GREs) or by interacting with various transcription factors. Alternatively, hormone-bound GR can repress gene transcriptions by blocking the nuclear translocation of transcription factors. GR is composed of a modulating domain and a DNA-binding domain at its N-terminal, and a ligand-binding domain at the C-terminal.

Specificity

Expected to react with all 10 spliced isoforms reported by UniProt (P04150-1 through P04150-10).

Immunogen

Epitope: Modulating region
KLH-conjugated linear peptide corresponding to a sequence from the modulating region of human glucocorticoid receptor.

Application

Anti-Glucocorticoid Receptor Antibody is an antibody against Glucocorticoid Receptor for use in Western Blotting, Affinity Binding Assay.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors
Western Blotting Analysis: A representative lot detected glucocorticoid receptor (GR) in mouse liver homogenates (Revollo, J.R., et al. (2013). Sci. Signal. 6(304):ra103).
Western Blotting Analysis: A representative lot detected both wild-type glucocorticoid receptor (GR) and the Dim4 DNA-binding mutant in lysates from U2OS cells that express the respective human GR constructs (Revollo, J.R., et al. (2013). Sci. Signal. 6(304):ra103).
Western Blotting Analysis: A representative lot detected both free glucocorticoid receptor (GR) and dexamethasone mesylate (Dex-Mes) labeled GR in HeLa S3 cell lysates (Cidlowski, J.A., et al. (1990). Mol. Endocrinol 4(10):1427-1437).
Affinity Binding Assay: A representative lot bound monomeric glucocorticoid receptor (GR) as well as oligomeric GR complexes in cytosolic preparations from dexamethasone-/Dex-treated HeLa S3 cells without affecting GR DNA-binding activity (Cidlowski, J.A., et al. (1990). Mol. Endocrinol 4(10):1427-1437).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected glucocorticoid receptor in 10 µg of HeLa cell lysate.

Target description

~95 kDa observed. 85.7 kDa (Alpha; Alpha-A), 81.5 kDa (Beta; Beta-A), 85.8 kDa (Alpha-2; Gamma), 81.7 kDa (Beta-2), 64.8 kDa (GR-A alpha), 60.6 kDa (GR-A beta), 82.8 kDa (Alpha-B). 78.7 kDa (Beta-B) calculated.

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service