Skip to Content
Merck
All Photos(2)

Key Documents

ABE1359

Sigma-Aldrich

Anti-PHD finger protein 14 (PHF14)

from rabbit, purified by affinity chromatography

Synonym(s):

PHF14

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$550.00

$214.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$550.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$214.00


Estimated to ship on08 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PHF14(9678)

General description

PHD finger protein 14 (UniProt: O94880) is encoded by the PHF14 (also known as KIAA0783) gene (Gene ID: 9678) in human. PHF14 is a member of the PHD finger protein family that serves as a chromatin-binding protein. It contains four putative PHD fingers, two coiled-coil regions, and four zinc fingers. PHF14 is highly conserved from C. elegans to humans and is considered as a transcriptional regulator of target gene expression. PHF14 has been identified as a regulator of mesenchyme growth in embryonic tissues. Two isoforms of PHF14 have been described that are produced by alternative splicing. PHF14 interacts with histones via its PHD1 and PHD3 domains indicating its role in epigenetic regulation. It is involved in chromatin-mediated transcriptional regulation and chromatin dynamics. During cell division, PHF14 is shown to be directly bound to and co-localized with KIF4A to form a functional complex. PHF14 is shown to be overexpressed in lung cancer and this overexpression is associated with poor prognosis of non-small cell lung carcinoma. (Ref.: Zhang, L., et al. (2017). Oncotarget. 8(12); 19684-19698; Yang, B., et al. (2017). Sci Rep. 7; 39888).

Specificity

This rabbit polyclonal antibody detects PHD Finger Protein 14 in human cells. It targets an epitope within 16 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of human PHD finger protein 14.

Application

Anti-PHD finger protein 14 (PHF14) Antibody, Cat. No. ABE1359, is a highly specific rabbit polyclonal antibody that detects PHD finger protein 14 and has been tested for use in Immunocytochemistry and in Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected PHD finger protein 14 (PHF14) in HeLa cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PHD finger protein 14 (PHF14) in HEK293 cell lysate.

Target description

100.0 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service