Skip to Content
Merck
All Photos(2)

Key Documents

ABE114

Sigma-Aldrich

Anti-PRDM5 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

PR domain zinc finger protein 5, PR domain-containing protein 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$476.00

$476.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$476.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$476.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRDM5(11107)

General description

PRDM5 is a member of the PR-domain containing family of tumor suppressors. PRDM5 activates histone methyltransferase G9a and class I histone deacetylases to regulate transcription. PRDM5 typically functions as a transcriptional repressor but can also be an activator for endogenous target genes. PRDM5 has often been found to be silenced in human breast, ovarian and liver cancers and and is a candidate biomarker for certain cancers, such as gastrointestinal cancer. Additional studies have also linked the loss of PRDM5 function to the development and pathogenesis of cervical cancer.

Immunogen

Linear peptide corresponding to human PRDM5.

Application

Use Anti-PRDM5 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect PRDM5 also known as PR domain-containing protein 5.
Western Blot Analysis:1 µg/mL from a representative lot detected PRDM5 on 10 µg of HeLa cell lysate and HeLa nuclear extract.

Quality

Evaluated by Western Blot in PC3 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PRDM5 on 10 µg of PC3 cell lysate.

Target description

~75 kDa observed. Three isoforms at 73, 70, and 13 kDa have been described. Uncharacterized bands between 13 and 73 kDa may be observed in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service