Skip to Content
Merck
All Photos(1)

Key Documents

ABE1075

Sigma-Aldrich

Anti-WD repeat-containing protein 5 Antibody

serum, from rabbit

Synonym(s):

WD repeat-containing protein 5, BMP2-induced 3-kb gene protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$727.00

$727.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$727.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$727.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WDR5(11091)

General description

WD repeat-containing protein 5 (WDR5) or alternatively BMP2-induced 3kb gene protein, is encoded by the human gene named WDR5/BIG3 and is part of a family of WD-40 repeat proteins that appear to facilitate the formation of multi-protein complexes much like a sticky tape will bind a variety of components together. The WD repeat proteins are involved in a wide array of cellular functions and processes including cell cycle progression, homeobox gene activation, apoptosis, gene regulation and histone methylation. One of WDR5 best studied activities is as part of a multi-protein complex that facilitates the methylation of histones, particularly histone H3, through its interaction with MLL a histone methyltransferase. WDR5 is also expressed in developing bone cells including osteoblasts, chondrocytes, and marrow stromal cells and works to accelerate osteoblast differentiation and positively promotes Wnt signaling in developing bone tissues.

Immunogen

Linear peptide corresponding to human WD repeat containing protein 5.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-WD repeat-containing protein 5 antibody is validated for use in western blotting for the detection of WD repeat-containing protein 5.
Western Blotting Analysis: A representative lot from an independent laboratory detected WD repeat containing protein 5 in tested cell lysates. (Wysocka, J., et al. (2003). Genes Dev. 17(7):896-911.; Tyagi, S., et al. (2007). 27(1):107-119.; Jin, S., et al. (2010). J Clin Invest. 120(2):593-606.; Tyagi, S., et al. (2009). EMBO J. 28(20):3185-3195.; Diao, Y., et al. (2012). Cell Stem Cell. 11(2):231-241.; Schram, A. W., et al. (2013). PLoS One. 8(7):e70035.).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected WD repeat containing protein 5 in 10 µg of NIH/3T3 cell lysate.

Target description

~37 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service