Skip to Content
Merck
All Photos(1)

Key Documents

ABE1047

Sigma-Aldrich

Anti-NRF2a/GABPA Antibody

serum, from rabbit

Synonym(s):

GA-binding protein alpha chain, GABP subunit alpha, Nuclear respiratory factor 2 subunit alpha, Transcription factor E4TF1-60, NRF2a/GABPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$597.00

$597.00


Estimated to ship on08 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$597.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$597.00


Estimated to ship on08 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable
electrophoretic mobility shift assay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GABPA(2551)

General description

GA-binding protein alpha chain (UniProt Q06546; also known as GABP subunit alpha, GABPalpha, NRF-2, Nuclear respiratory factor 2 subunit alpha, Transcription factor E4TF1-60) is encoded by the GABPA (Also known as E4TF1A) gene (Gene ID 2551) in human. Initially identified through their interactions with cytochrome c and cytochrome oxidase promoters, GABPA (NRF-2) and the related nuclear respiratory factor NRF-1 target many nuclear genes essential for mitochondrial respiratory function, including the mitochondrial transcription specificity factors TFB1M and TFB2M. GABPA plays an anti-oxidant, pro-survival role in mitochondrial biogenesis process. GABPA is upregulated in response to oxidant exposure or increased cellular reactive oxygen species (ROS), where it mediates the transcription of mitochondrial transcription factor A (mtTFA) that is needed for mitochondrial DNA transcription.

Immunogen

Recombinant protein corresponding to human NRF2a/GABPA.

Application

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected GABPA (NRF-2alpha) occupancy at the TFB1M and TFB2M promoters by ChIP using HeLa chromatin preparations (Gleyzer, N., et al. (2005). Mol Cell Biol. 2005 Feb;25(4):1354-66).
Western Blotting Analysis: A representative lot detected GABPA (NRF-2alpha) in U2OS cell lysates (Gleyzer, N., and Scarpulla, R.C. (2011). J. Biol. Chem. 286(46):39715-39725; Gleyzer, N., and Scarpulla, R.C. (2013). J. Biol. Chem. 288(12):8004-8015).
Electrophoretic Mobility Shift Assay (EMSA) Analysis: A representative lot caused a supershift of GABPA (NRF-2alpha)-DNA complex in EMSA using recombinant GABPA or heparin agarose-purified HeLa nuclear extract and radiolabeled synthetic oligonucleotides corresponding to cytochrome oxidase subunit IV, hTFB1M, or hTFB2M promoter sequence with GABPA recognition sites (Gleyzer, N., et al. (2005). Mol Cell Biol. 2005 Feb;25(4):1354-66).
Electrophoretic Mobility Shift Assay (EMSA) Analysis: A representative lot caused a supershift of GABPA (NRF-2alpha)-DNA complex in EMSA using in vitro translated GABPA and radiolabeled cytochrome oxidase subunit IV promoter fragment containing tandem GABPA recognition sites (Vercauteren, K., et al. (2008). J Biol Chem. 283(18):12102-12111).
Immunoprecipitation Analysis: A representative lot immunoprecipitated GABPA (NRF-2alpha)-PRC complex from human embryonic kidney 293FT cell lysate (Vercauteren, K., et al. (2008). J Biol Chem. 283(18):12102-12111).
This Anti-NRF2a/GABPA Antibody is validated for use in Western Blotting, Chromatin Immunoprecipitation (ChIP), Electrophoretic Mobility Shift Assay for the detection of NRF2a/GABPA.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected NRF2a/GABPA in 10 µg of HeLa cell lysate.

Target description

~55 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tongwu Zhang et al.
Cancer research, 77(7), 1649-1661 (2017-01-22)
SDHD encodes subunit D of the succinate dehydrogenase complex, an integral membrane protein. Across cancer types, recurrent SDHD promoter mutations were reported to occur exclusively in melanomas, at a frequency of 4% to 5%. These mutations are predicted to disrupt
Andrew M McKinney et al.
Cell reports, 40(12), 111344-111344 (2022-09-22)
Telomerase activation counteracts senescence and telomere erosion caused by uncontrolled proliferation. Epidermal growth factor receptor (EGFR) amplification drives proliferation while telomerase reverse transcriptase promoter (TERTp) mutations underlie telomerase reactivation through recruitment of GA-binding protein (GABP). EGFR amplification and TERTp mutations
Johan O Paulsson et al.
Endocrine-related cancer, 27(5), 295-308 (2020-03-13)
Mutations in the miRNA enzyme gene DICER1 have been reported in several endocrine malignancies and is associated with the rare tumour-predisposing DICER1 syndrome. DICER1 mutations have been reported in subsets of follicular thyroid carcinoma (FTC), but the role of DICER1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service