Skip to Content
Merck
All Photos(4)

Key Documents

ABD77

Sigma-Aldrich

Anti-Dax1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor subfamily 0 group B member 1, DSS-AHC critical region on the X chromosome protein 1, Nuclear receptor DAX-1, AHC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$751.00

$751.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$751.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$751.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NR0B1(190)

General description

Dax-1 is an orphan nuclear receptor and is required for the normal development of the hypothalamic-pituitary-adrenal-gonadal axis formation. Defects in Dax-1 are the cause of X-linked adrenal hypoplasia congenital (XL-AHC); also known as X-linked Addison disease (AHX). Dax-1 also has a role in the development of the embryo and in the maintenance of embryonic stem cell pluripotency by acting as a negative regulator of Oct3/4 in ES cells.

Specificity

75% Homologogous to Mouse Dax1

Immunogen

KLH-conjugated linear peptide corresponding to the Region of Ligand binding of Human Dax1.

Application

Evaluated by Immunohistochemistry in Human and Rat adrenal gland tissue. Immunohistochemsitry Analysis: A 1:1000 dilution of this antibody detected Dax1 in Human and Rat glioblastoma adrenal gland tissue.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Dax1 antibody is validated for use in WB & IHC for the detection of the Dax1 protein.

Quality

Evaluated by Western Blot in a Dax1 overexpressing HEK293 transfected cell line. Western Blot Analysis: A 1:2000 dilution of this antibody detected Dax1 in a Dax1 overexpressing HEK293 transfected cell line.

Target description

~53 kDa observed

Linkage

Replaces: 04-445

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Dax1 overexpressing HEK293 transfected cell line

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service