Skip to Content
Merck
All Photos(3)

Key Documents

ABD119

Sigma-Aldrich

Anti-TLE1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Transducin-like enhancer protein 1, E(Sp1) homolog, Enhancer of split groucho-like protein 1, ESG1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$508.00

$508.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$508.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

$508.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TLE1(7088)

General description

Transducin-like enhancer protein 1 (UniProt Q04724; also known as E(Sp1) homolog, Enhancer of split groucho-like protein 1, ESG1) is encoded by the TLE1 gene (Gene ID 7088) in human. TLE proteins interact with HDACs as well as deacetylated histone tails and play an important role in transcription regulation in several signaling pathways, including Ras, Notch, and Wnt. TLE proteins contain a conserved N-terminal glutamine-rich (Q) domain (a.a. 1-131 of TLE1) that mediates TLEs tetramerization as well as TCF/LEF proteins binding, and is essential for chromatin oligomerization. This is followed by a variable central domain sub-divided into a glycine-prolin-rich (GP) region (a.a. 132-199 of TLE1), the CcN (protein kinase CK2/cell cycle-dependent kinase 2/nuclear localization sequence) domain (a.a. 200-268), a serine-proline-rich (SP) region (a.a. 200-268 and 269-450 of TLE1), and a C-terminal WD40 domain with 6 WD repeats (a.a. 470-767 of TLE1) that interact with multiple transcription factors. The GP region binds directly to HDAC1, the CcN region contains consensus CK2 and Cdc2 phosphorylation sites that are modified in a cell cycle-dependent manner, the SP region is phosphorylated by the Ser/Thr kinases HIPK2 and MAPK, which appear to negatively regulate transcription repression activity, and may also interact with basic helix-loop-helix transcription factors. The C-terminal WD40 domain is essential for chromatin condensation and, together with SP, interacts with bHLH proteins.

Specificity

This polyclonal antibody targets the CcN (protein kinase CK2/cell cycle-dependent kinase 2/nuclear localization sequence) domain of TLE1 and stained cell nuclei by fluorescent immunocytochemistry.

Immunogen

Epitope: CcN domain.
KLH-conjugated linear peptide corresponding to a sequence within the CcN domain of human TLE1.

Application

Immunocytochemistry Analysis: 1 µg/mL from a representative lot detected TLE1 in HeLa cells and H9 human stem cells.
Research Category
Stem Cell Research
Research Sub Category
Histone Modifying Proteins
This Anti-TLE1 Antibody is validated for use in Western Blotting, Immunocytochemistry for the detection of TLE1.

Quality

Evaluated by Western Blotting in H9 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antiobdy detected TLE1 in 10 µg of H9 human stem cell lysate.

Target description

~90 kDa observed. 83.20 kDa calculated.

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service