Skip to Content
Merck
All Photos(1)

Key Documents

ABC848

Sigma-Aldrich

Anti-Toso Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fas apoptotic inhibitory molecule 3, Regulator of Fas-induced apoptosis Toso, Toso

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$561.00

$561.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$561.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$561.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FAIM3(9214)

General description

Apoptosis is an important process by which normal tissue homeostasis and function are maintained. Toso (also known as Fas apoptotic inhibitory molecule 3, Regulator of Fas-induced apoptosis and FAIM3) is a novel regulator of Fas-induced apoptosis in lymphoid cells and when overexpressed in Jurkat cells as well as transgenic mice render these cells resistant to Fas-induced apoptosis but not to TRAIL-induced apoptosis. Furthermore, Toso was found to associate with FADD, suggesting that Toso functions by disrupting the formation of the death-inducing signaling complex.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to a 13 amino acid peptide near the C-terminus of human Toso.

Application

Anti-Toso Antibody is a highly specific rabbit polyclonal antibody, that targets Fas apoptotic inhibitory molecule 3 & has been tested in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in human lung tissue lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected Toso in human lung tissue lysate with and without blocking peptide.

Target description

~60 kDa observed.
Despite its predicted molecular weight of 43 kDa, Toso often migrates at 60 kDa in western blots.
Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service