Skip to Content
Merck
All Photos(3)

Documents

ABC844

Sigma-Aldrich

Anti-IEX1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Radiation-inducible immediate-early gene IEX-1, Differentiation-dependent gene 2 protein, Protein DIF-2, Immediate early protein GLY96, Immediate early response 3 protein, PACAP-responsive gene 1 protein, Protein PRG1, IEX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IER3(8870)

General description

Radiation-inducible immediate-early gene IEX-1, also known as Differentiation-dependent gene 2 protein (DIF-2) or PACAP-responsive gene 1 protein (Protein PRG1), and encoded by the gene name IER3 or DIF2 or IEX1 or PRG1, is a stress inducible gene that is induced by ionizing radiation, ultraviolet radiation, and a variety of growth factors, i.e., FAS and TNF-α. IEX1 is widely expressed in epithelial and endocrine tissues, as well as in vascular endothelium. IEX1 plays an important role in the regulation of cellular growth, cell death and oncogenesis and is precisely regulated by multiple transcription factors such as p53, NF-κB/rel, Sp1 and c-Myc, to ensure rapid and transient expression of IEX1 in cells under a variety of stress conditions. IEX1 is expressed as both a longer form (IEX1L) and a splice variant, designated IEX1S. It is localized to the nucleus and perinuclear region. Overexpression of IEX1 facilitates apoptosis and cell cycle progression, whereas disruption of IEX1 expression is associated with decreases in both apoptosis and cell cycle progression.

Immunogen

Epitope: Center region
KLH-conjugated synthetic peptide corresponding to the center region of human IEX1.

Application

Anti-IEX1 Antibody is an antibody against IEX1 for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 2.5 μg/mL from a representative lot detected IEX1 in human liver tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected IEX1 in human brain cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 4 μg/mL of this antibody detected IEX1 in human brain tissue lysate.

Target description

~16 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service