Skip to Content
Merck
All Photos(3)

Key Documents

ABC839

Sigma-Aldrich

Anti-ATR Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Anthrax toxin receptor 1, Tumor endothelial marker 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$561.00

$561.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$561.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$561.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANTXR1(84168)

General description

The Anthrax toxin receptor 1 (also known as Tumor endothelial marker 8, ANTXR1 and ATR) was initially discovered as the tumor endothelial marker (TEM8). This protein, which exists in three isoforms (33, 37, 41, and 63 kDa), is highly expressed in tumor vessels as well as in the vasculature of developing embryos, suggesting that it may normally play a role in angiogenesis. However, it also acts as the receptor for anthrax toxin. Following the binding of this protein by the protective antigen (PA) of anthrax, PA is cleaved and heptamerizes to form the binding site for both edema factor (EF) and lethal factor (LF). This complex is then endocytosed by the cell; acidification in endosomes allows the release of EF and LF into the cytoplasm where they interfere with MAPK signaling and induce apoptosis.

Immunogen

Epitope: Center region
KLH-conjugated linear peptide corresponding to 13 amino acids near the center region of human ATR.

Application

Anti-ATR Antibody is a highly specific rabbit polyclonal antibody, that targets ATR & has been tested in western blotting, IHC & ICC.
Immunohistochemistry Analysis: 2 μg/mL from a representative lot detected ATR in human brain tissue.

Immunocytochemistry Analysis: 10 μg/mL from a representative lot detected ATR in human brain cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2 μg/mL of this antibody detected ATR in HepG2 cell lysate.

Target description

~41 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified by Ion-Exchange Chromatography
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service