Skip to Content
Merck
All Photos(2)

Key Documents

ABC457

Sigma-Aldrich

Anti-dUTPase Antibody

from rabbit

Synonym(s):

Deoxyuridine 5′-triphosphate nucleotidohydrolase, mitochondrial, dUTPase, dUTP pyrophosphatase, dUTPase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$660.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DUT(1854)

General description

dUTPase, also known as dUTP pyrophosphatase, and encoded by the gene DUT, is a critical enzyme involved in nucleotide metabolism for the production of dUMP, the immediate precursor of thymidine nucleotides. dUTPase normally can also function to decrease the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA and cause mutations. dUTPase is localized to the nucleus and mitochondrial locations. dUTPase is expressed in numerous tissues and has multiple isoforms. Some isoforms are constitutively expressed while others match DNA replication cycles. Because dUTPase plays a pivotal role in regulating cellular dUTP pools, this enzyme could have profound effects on the efficacy of agents that target thymidylate biosynthesis and dUTPase expression is being examined as a prognostic marker for overall survival and response to therapy in colon and other cancers where thymidylate therapy is used.

Immunogen

Recombinant protein corresponding to human dUTPase.

Application

Anti-dUTPase Antibody is a highly specific rabbit polyclonal antibody, that targets dUTPase & has been tested in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 2 µg/mL from a representative lot detected dUTPase in 10 µg of A431, HEK293, and HeLa cell lysates and human placenta tissue lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected dUTPase in MDA-MB231, MCF-7, H184B5F5/M10 cell lysate (Hu, C. M., et al. (2012). Cancer Cell. 22(1):36-50.).

Quality

Evaluated by Western Blotting in MDA-MB-231 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected dUTPase in 10 µg of MDA-MB-231 cell lysate.

Target description

~18 kDa observed. The expected molecular weight of this protein is 27 kDa However, this protein has been observed at 18 kDa (Prof. Z. F. Chang, National Yang-Ming University). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service