Skip to Content
Merck
All Photos(1)

Key Documents

ABC442

Sigma-Aldrich

Anti-Cathepsin Z Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cathepsin Z, Cathepsin P, Cathepsin X

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$683.00

$683.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTSZ(1522)

General description

Cathepsin Z, also known as Cathepsin P, Cathepsin X, and encoded by the gene name CTSZ, is a member of the the peptidase C1 family. Cathepsin Z is a lysosomal cysteine proteinase that exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. Cathepsin Z is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis. At least two transcript variants of this gene have been found, but the full-length nature of only one of them has been determined.

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human Cathepsin Z near the C-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Cathepsin Z antibody is validated for use in Western Blotting for the detection of Cathepsin Z.

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Cathepsin Z in 10 µg of human liver tissue lysate.

Target description

~33 kDa observed. (Pungecar, J., et al. (2000). PMID: 10653163)

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service