Skip to Content
Merck
All Photos(2)

Key Documents

ABC335

Sigma-Aldrich

Anti-MTCH2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

MTCH2, Mitochondrial carrier homolog 2, HSPC032, MIMP, Met-induced mitochondrial protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$584.00

$584.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$584.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$584.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTCH2(23788)

General description

Mitochondrial carrier homolog 2 (MTCH2) may play a key role in the mitochondrial apoptosis pathway and may induce mitochondrial depolarization. MTCH2 is suggested to catalyze the exchange of substrates across the inner mitochondrial membrane in response to apoptotic signals. Additionally, it is suggested to be targeted by Bid, a pro-apoptotic Bcl-2 family member. The Bcl-2 family of proteins incorporates a number of critical regulators of apoptosis that can be divided into two classes: inhibitors of apoptosis and promoters of cell death. Apoptosis plays a major role in tissue homeostasis, normal organism development, and removal of damaged cells. The disruption of this process has been linked to a variety of diseases such as cancer.

Immunogen

KLH-conjugated linear peptide corresponding to human MTCH2.

Application

Anti-MTCH2 Antibody is a highly specific rabbit polyclonal antibody, that targets Mitochondrial carrier homolog 2 & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected MTCH2 in mouse kidney tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected MTCH2 in mouse kidney cells.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected MTCH2 in 15 µg of HEK293 cell lysate.

Target description

~34 kDa observed.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service