Skip to Content
Merck
All Photos(2)

Key Documents

ABC212

Sigma-Aldrich

Anti-phospho ULK1 Antibody (Thr574)

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein kinase ULK1, Serine/threonine-protein kinase Unc51.1, Unc-51-like kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr574)

Gene Information

human ... ULK1(8408)

General description

ULK1 or autophagy-related protein 1 homolog (ATG1) is a serine/threonine protein kinase. It is a ubiquitously expressed protein that is highly abundant in the brain, skeletal muscle, and heart. It is involved in the signaling pathway of mTOR receptors leading to the regulation of autophagy in nutrient-deprived cells. mTOR may interact with ULK1 as a complex of ULK1/FIP200/Atg13. ULK1 may play an important role in neuronal development: Elimination of ULK1′s enzymatic activity by targeted disruption of its kinase domain has blocked the outgrowth of neurons in the developing mouse brain, possibly by blocking the ULK1-mediated trafficking of neurotrophins.

Immunogen

KLH-conjugated linear peptide corresponding to mouse ULK1 phosphorylated at Thr574.

Application

Detect Autophagy-related protein;ATG1 using this rabbit polyclonal antibody, Anti-phospho ULK1 Antibody (Thr574) validated for use in western blotting & Peptide Inhibition Assay.
Western Blotting/Peptide Inhibition Analysis: 0.5 µg/mL from a representative detected phospho ULK1 in WB/PIA.

Quality

Evaluated by Western Blotting in untreated and Calyculin A with okadaic acid treated Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected phospho ULK1 (Thr574) in 10 µg of Jurkat cell lysate.

Target description

~120 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service