Skip to Content
Merck
All Photos(2)

Key Documents

ABC156

Sigma-Aldrich

Anti-BAG family molecular chaperone regulator 3 (BAG3) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Bcl-2-associated athanogene 3, Bcl-2-binding protein Bis, Docking protein CAIR-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$552.00

$552.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$552.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$552.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BAG3(9531)

General description

Bcl2-associated athanogene 3 (BAG3), also known as CAIR-1 or Bis, belongs to a family of six evolutionarily conserved co-chaperone proteins that contain a characteristic C-terminal BAG domain. In most cell types, BAG3 is localized to the cytoplasm, where it regulates the heat shock protein 70 chaperone. BAG3 is activated in response to stressful stimuli, and plays a critical role in modulating multiple cellular processes, including apoptosis, autophagy, embryonic development, and actin rearrangement. Abnormal expression of BAG3 results cardiomyopathy dilated type 1HH.

Immunogen

KLH-conjugated linear peptide corresponding to human BAG3.

Application

Detect BAG family molecular chaperone regulator 3 (BAG3) using this Anti-BAG family molecular chaperone regulator 3 (BAG3) Antibody validated for use in Western Blotting, IHC(P).
Immunohistochemistry Analysis: A 1:500-1:1,000 dilution from a representative lot detected BAG3 in human cardiac and renal tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in DU145 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected BAG3 in 10 µg of DU145 cell lysate.

Target description

~74/40 kDa observed. An uncharacterized band may appear at ~65 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
DU145 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service