Skip to Content
Merck
All Photos(1)

Key Documents

ABC1414

Sigma-Aldrich

Anti-phospho IGPR-1 (Ser220)

Synonym(s):

Transmembrane and immunoglobulin domain-containing protein 2;CD28 homolog;Immunoglobulin and proline-rich receptor 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$264.00
100 μG
$665.00

$264.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$264.00
100 μG
$665.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$264.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

mol wt

calculated mol wt 30.68 kDa
observed mol wt ~45 to 55 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

target post-translational modification

phosphorylation (pSer220)

Gene Information

human ... TMIGD2(126259)

General description

Transmembrane and immunoglobulin domain-containing protein 2 (UniProt: Q96BF3; also known as CD28 homolog, Immunoglobulin and proline-rich receptor 1, IGPR-1) is encoded by the TMIGD2 (also known as CD28H, IGPR1, UNQ3059/PRO9879) gene (Gene ID: 126259) in human. IGPR-1 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 23-150), a transmembrane domain (aa 151-171), and a cytoplasmic domain (aa 172-282). It is widely expressed with higher expression observed in epithelial and endothelial cells, including bronchial epithelial cells of lung, breast glandular and lobular epithelia cells. It plays a role in cell-cell interaction, cell migration, and angiogenesis. IGPR-1 is localized to endothelial adherens junctions, and through trans-homophilic dimerization it regulates endothelial cell-cell adhesion and barrier function. This dimerization stimulates phosphorylation of serine 220, which is required for IGPR-1 to regulate endothelial barrier function and angiogenesis. Through interaction with HHLA2, IGPR-1 is reported to co-stimulate T-cells in the context of TCR-mediated activation and enhances T-cell proliferation and cytokine production via an Akt-dependent signaling cascade. IGPR-1 is also phosphorylated on serine 220 by I B kinase β, which leads to its activation. Activated IGPR-1 then stimulates phosphorylation of AMPK, which in turn phosphorylates ULK1 and Beclin-1 and leads to increased levels of LC3-II. Genotoxic agents such as doxorubicin and flow shear stress are also reported to simulate phosphorylation of IGPR-1 at serine 220. (Ref.: Amraei, R., et al. (2020). J. Biol. Chem. 295(49); 16691-16699; Wang, YHW., et al.(2016). J. Mol. Biol. 428(24; PtB); 5019-5033).

Specificity

This rabbit polyclonal antibody detects Immunoglobulin and proline-rich receptor 1 (IGPR-1) phosphorylated on serine 220.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids surrounding phosphoserine 220 in the cytoplasmic domain in Human Immunoglobulin and proline-rich receptor 1 (IGPR-1).

Application

Qualtiy Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells overexpressing IGPR-1.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected IGPR-1 phosphorylated on serine 220 in lysates from HEK293 cells overexpressing IGPR-1.

Tested applications

Immunofluorescence Analysis: A representative lot detected phospho-IGPR-1 (Ser220) in Immunofluorescence applications (Wang, Y., et. al. (2016). J Mol Biol. 428(24 Pt B):5019-5033).

Immunohistochemistry Applications: A representative lot detected phospho-IGPR-1 (Ser220) in Immunohistochemistry applications (Wang, Y., et. al. (2016). J Mol Biol. 428(24 Pt B):5019-5033).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified rabbit polyclonal antibody in PBS without azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service