Skip to Content
Merck
All Photos(1)

Key Documents

AB9704

Sigma-Aldrich

Anti-LRRK2 Antibody

serum, Chemicon®

Synonym(s):

Dardarin, Leucine-rich repeat kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$602.00

$602.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$602.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$602.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LRRK2(120892)

Specificity

Recognizes LRRK2 (leucine-rich repeat kinase 2, Dardarin)

Immunogen

Synthetic peptide from human LRRK2.

Application

Detect LRRK2 using this ANTI-LRRK2 Antibody validated for use in IH.
Immunohistochemistry: 1:1,000-1:8,000. The antibody works well on both cryosections and paraffin embedded tissues.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

Sheep serum. Lyophilized. Reconstitute with 100 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C to -70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (ACS grade or better) can be added (1:1) for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saskia Biskup et al.
BMC neuroscience, 8, 102-102 (2007-11-30)
Mutations within the leucine-rich repeat kinase 2 (LRRK2) gene account for a significant proportion of autosomal-dominant and some late-onset sporadic Parkinson's disease. Elucidation of LRRK2 protein function in health and disease provides an opportunity for deciphering molecular pathways important in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service