Skip to Content
Merck
All Photos(1)

Key Documents

AB9600

Sigma-Aldrich

Anti-Neuropilin-1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$617.00

$617.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$617.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$617.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NRP1(8829)

Specificity

It is expected that the antibody will also react with rat and mouse.
Neuropilin-1. The immunogen sequence has low homology with neuropilin-2.

Immunogen

Synthetic peptide from the N-terminal CUB domain of human neuropilin-1.

Application

Anti-Neuropilin-1 Antibody is an antibody against Neuropilin-1 for use in WB.
Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance
Western blot: 1:500 on transfected COS-7 cells. The antibody reacts with the ~130 kDa
protein. The suggested antibody diluent is PBS containing 5% non-fat milk and 0.04%
Tween 20.

Physical form

Affinity purified immunoglobulin in PBS containing 50% glycerol, 1 mg/mL BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srinivas R Viswanathan et al.
Nature genetics, 50(7), 937-943 (2018-06-30)
Functional redundancy shared by paralog genes may afford protection against genetic perturbations, but it can also result in genetic vulnerabilities due to mutual interdependency1-5. Here, we surveyed genome-scale short hairpin RNA and CRISPR screening data on hundreds of cancer cell
Stefanie Rauskolb et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(5), 1739-1749 (2010-02-05)
Although brain-derived neurotrophic factor (BDNF) is linked with an increasing number of conditions causing brain dysfunction, its role in the postnatal CNS has remained difficult to assess. This is because the bdnf-null mutation causes the death of the animals before

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service