Skip to Content
Merck
All Photos(2)

Key Documents

AB951-I

Sigma-Aldrich

Anti-Neuron Specific Enolase Antibody

from rabbit

Synonym(s):

Gamma-enolase, 2-phospho-D-glycerate hydro-lyase, Enolase 2, Neural enolase, Neuron-specific enolase, NSE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$833.00

$833.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$833.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$833.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ENO2(2026)

General description

The glycolytic enzyme enolase (2-phospho-D-glycerate hydrolase, E.C. 4.2.1.11) is composed of two subunits and exists in three molecular forms in human brain: non-neuronal enolase (NNE; alpha, alpha′-enolase), neuron-specific enolase (NSE; gamma,gamma′-enolase, also known as 14-3-2 protein and NSP) and the short-lived heterodimer alpha, gamma′-enolase. The neuron specific form is a highly acidic homodimeric protin of 78kDa, which has been found in central and peripheral nervous elements and also in many neuroendocrine cells and derivative tumors. NSE is regarded as the best described serum marker for small cell lung cancer. In addition, the measurement of NSE levels in both serum and cerebrospinal fluid following cerebral ischaemia and traumatic head injury provides a reliable laboratory indicator of the degree of brain cell damage and may allow early prediction of prognostic outcome.

Specificity

This antibody recognizes a region near the N-terminus of Neuron Specific Enolase.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to the near N-terminus of human Neuron Specific Enolase.

Application

Anti-Neuron Specific Enolase Antibody is a highly specific rabbit polyclonal antibody, that targets Neuron-Specific Enolase & has been tested in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected in Neuron Specific Enolase in human thalamus tissue and glioma sections.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Neuron Specific Enolase in 10 µg of rat brain tissue lysate.

Target description

~47 kDa observed. Cat. No. AB951, Anti-Neuron Specific Enolase detects Alpha Enolase (ENO1) at ~39kDa However, Neuron Specific Enolase has been observed at ~48 kDa (Braus, B., K., et al (2008). Vet Immunol Immunopathol 124(1-2):177-183.; Feo, S., et al. (2000). FEBS Lett. 473(1):47-52.).

Linkage

Replaces: AB951

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin H Quinn et al.
International journal of bioprinting, 9(4), 723-723 (2023-06-16)
The use of three-dimensional (3D) bioprinting has remained at the forefront of tissue engineering and has recently been employed for generating bioprinted solid tumors to be used as cancer models to test therapeutics. In pediatrics, neural crest-derived tumors are the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service