Skip to Content
Merck
All Photos(1)

Key Documents

AB8230

Sigma-Aldrich

Anti-Syndecan-1 Antibody

serum, from rabbit

Synonym(s):

CD138 antigen, heparan sulfate proteoglycan fibroblast growth factor

receptor, syndecan 1, syndecan proteoglycan 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$674.00

$674.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$674.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$674.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SDC1(6382)

General description

Syndecan-1, also known as CD138, is a transmembrane (Type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization. Bearing properties of both heparan sulfate and chondroitin sulfate, Syndecan-1 functions as a cell surface proteoglycan membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Altered syndecan-1 expression has been detected in several different tumor types.

Specificity

Cat. No. AB8230 recognizes mouse Syndecan-1.
Predicted to cross react with human, based on 100% sequence homology.
Reactivity with other species has not been tested.

Immunogen

His-tagged, full-length human recombinant Syndecan-1 protein.

Application

Anti-Syndecan-1 Antibody is an antibody against Syndecan-1 for use in WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated on a representative lot by Western blot on mouse liver lysate using 1:1000 dilution of Anti-Syndecan-1.

Target description

The expected molecular weight of monomeric Syndecan-1 is ~32 kDa; however, the core protein often migrates as an SDS-stable dimer with an approximate M.W. of 77 kDa Additionally, a high M.W. form of the protein may often be seen between 500-1000 kDa, likely the mature form of Syndecan-1, with all of its heparan and chondroitin sulfate additions. In whole tissues, this mature form often binds tightly to ECM molecules and may require solublization by urea extraction for additional SDS-PAGE analysis (Wilsie, 2006). Triton X-100 or similar mild detergents are sufficient to release the immature precursor forms of Syndecan-1.

Physical form

Rabbit serum containing 0.05% NaN3.

Storage and Stability

Maintain at -20°C for up to 1 year from date of receipt.

Analysis Note

Control
Mouse liver lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service