Skip to Content
Merck
All Photos(2)

Key Documents

AB5932

Sigma-Aldrich

Anti-Potassium Channel KvLQT1 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1, KQT-like 1, Voltage-gated potassium channel subunit Kv7.1, kidney and cardiac voltage dependent K+ channel, potassium voltage-gated channel, KQT-like subfamily, member 1, slow delay

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$757.00

$757.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$757.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$757.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNQ1(3784)

General description

KvLQT1(Kv7.1) is a voltage-gated potassium channel protein coded for by the gene KCNQ1. This protein is present in the cell membranes of cardiac muscle tissue and in inner ear neurons among other tissues. KvLQT1 can form heteromultimers with two other potassium channel proteins, KCNE1 and KCNE3 and in heart cells is required for the repolarization phase of the cardiac action potential. Mutations in the gene are associated with hereditary long QT syndrome, Romano-Ward syndrome, Jervell and Lange-Nielsen syndrome and familial atrial fibrillation, lead to a defective KvLQT1 protein and several forms of inherited arrhythmias known as Long QT syndrome, Short QT syndrome, and Familial Atrial Fibrillation.

Specificity

This antibody recognizes Potassium Channel KvLQT1 protein.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide from the C-terminus of human KvLQT1.

Application

Anti-Potassium Channel KvLQT1 Antibody, C-terminus detects level of Potassium Channel KvLQT1 & has been published & validated for use in IH & WB.
Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected Potassium Channel KvLQT1 in human myocardium tissue.

/Western Blotting Analysis: 2 μg/mL of this
antibody detected KvLQT1 in 10 μg of human
heart tissue lysate.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Evaluated by Western Blot in human heart tissue lysate.

Western Blot Analysis: 2 µg/ml of this antibody detected KvLQT1 in 10 µg of human heart tissue lysate.

Target description

~ 72 kDa Observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human heart tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fu Liang Ng et al.
British journal of pharmacology, 162(1), 42-53 (2010-09-16)
KCNQ-encoded voltage-gated potassium channels (K(v) 7) have recently been identified as important anti-constrictor elements in rodent blood vessels but the role of these channels and the effects of their modulation in human arteries remain unknown. Here, we have assessed KCNQ
Preet S Chadha et al.
British journal of pharmacology, 166(4), 1377-1387 (2012-01-19)
The aim of this study was to characterize the functional impact of KCNQ1-encoded voltage-dependent potassium channels (K(v)7.1) in the vasculature. Mesenteric arteries, intrapulmonary arteries and thoracic aortae were isolated from adult rats. K(v)7.1 channel expression was established by fluorescence immunocytochemistry.
Structural interplay of KV7.1 and KCNE1 is essential for normal repolarization and is compromised in short QT syndrome 2 (KV7.1-A287T).
Ina Rothenberg et al.
HeartRhythm case reports, 2(6), 521-529 (2017-05-12)
The KCNE2 potassium channel ancillary subunit is essential for gastric acid secretion.
Roepke, TK; Anantharam, A; Kirchhoff, P; Busque, SM; Young, JB; Geibel, JP; Lerner, DJ; Abbott, GW
The Journal of Biological Chemistry null
Lipopolysaccharide-induced inflammation attenuates taste progenitor cell proliferation and shortens the life span of taste bud cells.
Cohn, ZJ; Kim, A; Huang, L; Brand, J; Wang, H
BMC Neuroscience null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service