Skip to Content
Merck
All Photos(1)

Key Documents

AB5820

Sigma-Aldrich

Anti-Syntaxin 1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STX1A(6804)

Specificity

Recognizes Syntaxin 1A and Syntaxin 1B. The epitope does not share homology with any other known proteins.

Immunogen

GST fusion protein corresponding to a cytoplasmic part of rat Syntaxin 1A (Accession number P32851) (Bennet et al. 1992; Inoue & Akagawa 1992).

Application

Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This Anti-Syntaxin 1 Antibody is validated for use in WB, IH for the detection of Syntaxin 1.
Western blot: 1:700-1:1,250 (0.8-1.5 g/mL) using ECL on rat brain membranes.

Immunohistochemistry: AB5820 is directed against an intracellular epitope. Thus, a procedure including permeabilization of cells with 0.2% Triton X 100 is recommended. It is recommended that you start with a working concentration of 30 μg/mL (1:33).

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 50 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 mL of PBS. For positive control, in Western blot using 20 ng of protein per Minigel lane. For negative control, preincubate 5 μg of fusion protein with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam D Bachstetter et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(14), 6143-6153 (2013-04-05)
Neuropathology after traumatic brain injury (TBI) is the result of both the immediate impact injury and secondary injury mechanisms. Unresolved post-traumatic glial activation is a secondary injury mechanism that contributes to a chronic state of neuroinflammation in both animal models
José L Moreno et al.
Science signaling, 9(410), ra5-ra5 (2016-01-14)
Heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors (GPCRs) can form multiprotein complexes (heteromers), which can alter the pharmacology and functions of the constituent receptors. Previous findings demonstrated that the Gq/11-coupled serotonin 5-HT2A receptor and the Gi/o-coupled metabotropic glutamate 2 (mGlu2)
Suleman Hussain et al.
Frontiers in molecular neuroscience, 9, 10-10 (2016-02-24)
Syntaxins are a family of membrane-integrated proteins that are instrumental in exocytosis of vesicles. Syntaxin-1 is an essential component of the presynaptic exocytotic fusion machinery in the brain and interacts with several other proteins. Syntaxin-1 forms a four-helical bundle complex
Nancy J Alexander et al.
Alcoholism, clinical and experimental research, 42(9), 1661-1673 (2018-06-27)
Recent work with long-term ethanol (EtOH) self-administration in nonhuman primate models has revealed a complex array of behavioral and physiological effects that closely mimic human alcohol abuse. Detailed neurophysiological analysis in these models suggests a myriad of pre- and postsynaptic
Dominic A Gioia et al.
Frontiers in neuroscience, 11, 424-424 (2017-08-09)
Chronic exposure to alcohol produces adaptations within the basolateral amygdala (BLA) that are associated with the development of anxiety-like behaviors during withdrawal. In part, these adaptations are mediated by plasticity in glutamatergic synapses occurring through an AMPA receptor mediated form

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service