Skip to Content
Merck
All Photos(1)

Key Documents

AB5718

Sigma-Aldrich

Anti-Hey L/HRT3 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HEYL(26508)

Specificity

Recognizes Hey L / HRT3, a Helix-loop-helix class of transcription factor.

Immunogen

Synthetic peptide, amino acids 34-47 of human Hey L / HRT3.

Application

Anti-Hey L/HRT3 Antibody detects level of Hey L/HRT3 & has been published & validated for use in WB, IH.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naresh K Rajendran et al.
Alcoholism, clinical and experimental research, 45(11), 2217-2230 (2021-09-30)
Arterial endothelium plays a critical role in maintaining vessel homeostasis and preventing atherosclerotic cardiovascular disease (CVD). Low-to-moderate alcohol (EtOH) consumption is associated with reduced atherosclerosis and stimulates Notch signaling in endothelial cells. The aim of this study was to determine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service