Skip to Content
Merck
All Photos(1)

Key Documents

AB5699

Sigma-Aldrich

Anti-Cysteine-rich Motor Neuron 1 Antibody

serum, Chemicon®

Synonym(s):

CRIM1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$898.00

$898.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$898.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$898.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CRIM1(51232)

Specificity

Crim1

SUBCELLULAR LOCALIZATION: Intracellular and Membrane associated.

Immunogen

Recombinant protein from the cytoplasmic tail of mouse Crim1.

Application

Anti-Cysteine-rich Motor Neuron 1 Antibody detects level of Cysteine-rich Motor Neuron 1 & has been published & validated for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
Western blot: 1:1,000 using ECL. The antibody reacts with proteins of ~120 kDa. Preferred gel percentage is 7.5%. Suggested blocking buffer is PBS containing 5% non-fat milk. Suggested dilution buffer is PBS containing 1% non-fat milk.

Immunoprecipitation: 5 μL to 1 mL of cell extract. Preferred tissue/cell lysis buffer is RIPA. Suggested incubation time is 2-4 hours. Suggested capture agent is Protein A.

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingxian Lu et al.
Investigative ophthalmology & visual science, 61(12), 21-21 (2020-10-22)
Yap1 encodes an evolutionarily conserved transcriptional coactivator and functions as a down-stream effector of the Hippo signaling pathway that controls tissue size and cell growth. Yap1 contributes to lens epithelial development. However, the effect of Yap1 haplodeficiency on the lens

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service