Skip to Content
Merck
All Photos(1)

Key Documents

AB5509

Sigma-Aldrich

Anti-Opioid Receptor µ Antibody

Chemicon®, from guinea pig

Synonym(s):

MOR-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$839.00

$839.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
50 μL
$839.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$839.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

guinea pig

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

primate, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... OPRM1(4988)

Specificity

Mu Opioid receptor.

Immunogen

Synthetic peptide (Catalog number AG375) from the C-terminus of rat mu Opioid receptor corresponding to residues 384-398 of the rat mu opioid receptor.

Application

Anti-Opioid Receptor μ Antibody-AB5509 is an antibody against Opioid Receptor for use in IC, IH.
Immunohistochemistry: 1:50-500 See Protocol.

Immunocytochemistry: 1:50-500 See Protocol.

Optimal working dilutions must be determined by the end user.

IHC Protocol: Male Sprague-Dawley rats (b.wt. 100-150g) were anesthetized with sodium pentobarbital and perfused via the ascending aorta with: 1) 50 mL of Ca2+-free Tyrode+s solution followed by 2) a paraformaldehyde-picric acid fixative and 3) 10% sucrose in PBS as a cryo-protectant. Tissues were rapidly dissected out and stored overnight in 0.1 M phosphate buffer (pH 7.4) containing 10% sucrose. Slide-mounted tissue sections were incubated with blocking buffer for 1 hour at room temperature. Primary antibody was diluted in blocking buffer to the appropriate working dilution. Blocking buffer was removed and the slides were then incubated at 2-8°C for 18-24 hours with AB5509. After rinsing in PBS 3 times sections were incubated for 60 minutes at room temperature with Cy3-conjugated secondary antibodies. After mounting in a mixture of PBS and glycerol (1:3) containing 0.1% p-phenylenediamine, sections were examined with a Nikon Microphot-SA epifluorescence microscope.

ICC Protocol: Mu opioid receptor transfected cells were processed for indirect immunofluorescence. Media was removed and cells were gently washed 3 times with serum free media. Media was removed and cells were gently washed 3 times with serum free media. Following fixation, cells were processed for indirect immunofluorescence as described above.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Linkage

Replaces: AB1774

Physical form

PBS containing 50% glycerol; Concentration 1.0 mg/ml

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annelyn Torres-Reverón et al.
Reproductive sciences (Thousand Oaks, Calif.), 23(9), 1158-1167 (2016-04-20)
Studies have examined how endometriosis interacts with the nervous system, but little attention has been paid to opioidergic systems, which are relevant to pain signaling. We used the autotransplantation rat model of endometriosis and allowed to progress for 60 days.
D Gagnon et al.
Scientific reports, 7, 41432-41432 (2017-01-28)
The loss of nigrostriatal dopamine neurons in Parkinson's disease induces a reduction in the number of dendritic spines on medium spiny neurons (MSNs) of the striatum expressing D1 or D2 dopamine receptor. Consequences on MSNs expressing both receptors (D1/D2 MSNs)
Allain-Thibeault Ferhat et al.
Frontiers in molecular neuroscience, 16, 1139118-1139118 (2023-04-04)
Autism is characterized by atypical social communication and stereotyped behaviors. Mutations in the gene encoding the synaptic scaffolding protein SHANK3 are detected in 1-2% of patients with autism and intellectual disability, but the mechanisms underpinning the symptoms remain largely unknown.
Yong-Hui Liao et al.
Molecular medicine reports, 17(2), 2803-2810 (2017-12-20)
Chronic pain occurs in ~85-90% of chronic pancreatitis (CP) patients. However, as the pathogenesis of CP pain remains to be fully understood, the current therapies for CP pain remain inadequate. Emerging evidence has suggested that the epigenetic modulations of genes
Esraa Mohamed et al.
Glia, 68(7), 1513-1530 (2020-02-18)
The generation of fully functional oligodendrocytes, the myelinating cells of the central nervous system, is preceded by a complex maturational process. We previously showed that the timing of oligodendrocyte differentiation and rat brain myelination were altered by perinatal exposure to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service