Skip to Content
Merck
All Photos(1)

Key Documents

AB5495

Sigma-Aldrich

Anti-Potassium Channel Kir6.2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$1,080.00

$1,080.00


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$1,080.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$1,080.00


Estimated to ship on25 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNJ11(3767)

Specificity

Recognizes Kir6.2 (Inward rectifier potassium channel Kir6.2). The antibody does not react with Kir6.1. The epitope specific for Kir6.2 is not present in any other known proteins.

Immunogen

Highly purified peptide corresponding to residues 372-385 of rat Kir6.2 (Accession P70673). The immunogen sequence is identical in mouse and conserved in human (12/14).

Application

Anti-Potassium Channel Kir6.2 Antibody detects level of Potassium Channel Kir6.2 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blotting: 1:200 using ECL on rat pancreas lysate.

Immunohistochemistry on rat pancrease sections. Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.025% sodium azide. Reconstitute with 50 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria S Lemak et al.
Frontiers in cellular neuroscience, 8, 255-255 (2014-09-16)
Medial temporal lobe structures are essential for memory formation which is associated with coherent network oscillations. During ontogenesis, these highly organized patterns develop from distinct, less synchronized forms of network activity. This maturation process goes along with marked changes in
Andrew G Edwards et al.
The Journal of physiology, 587(Pt 23), 5723-5737 (2009-10-07)
The female myocardium, relative to that of the male, exhibits sustained resistance to ischaemic tissue injury, a phenomenon termed sex-specific cardioprotection (SSC). SSC is dependent upon the sarcolemmal K(ATP) channel (sarcK(ATP)), and protein kinase C (PKC). Here we investigate whether
Chisayo Kozuka et al.
British journal of pharmacology, 172(18), 4519-4534 (2015-07-04)
γ-Oryzanol, derived from unrefined rice, attenuated the preference for dietary fat in mice, by decreasing hypothalamic endoplasmic reticulum stress. However, no peripheral mechanisms, whereby γ-oryzanol could ameliorate glucose dyshomeostasis were explored. Dopamine D2 receptor signalling locally attenuates insulin secretion in
Atsuko Miura et al.
The Journal of biological chemistry, 281(8), 5246-5257 (2005-12-27)
Mutations in the hepatocyte nuclear factor (HNF)-4alpha gene cause a form of maturity-onset diabetes of the young (MODY1) that is characterized by impairment of glucose-stimulated insulin secretion by pancreatic beta-cells. HNF-4alpha, a transcription factor belonging to the nuclear receptor superfamily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service