Skip to Content
Merck
All Photos(1)

Key Documents

AB5493

Sigma-Aldrich

Anti-Sodium Channel Nav1.5 Antibody, pain

Chemicon®, from rabbit

Synonym(s):

SCN5A

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$567.00

$567.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
$567.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$567.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SCN5A(6331)

Specificity

Recognizes Nav1.5 (Scn5a, SKM2). The epitope specific for Nav1.5 is not present in any other known proteins.

SPECIES REACTIVITIES:
The immunogen sequence is identical in mouse and conserved in human (17/19). Reactivity with other species has not yet been tested.

Immunogen

Highly purified peptide corresponding to residues 493-511 of rat Nav1.5 (Accession P15389).

Application

Anti-Sodium Channel Nav1.5 Antibody, pain is an antibody against Sodium Channel Nav1.5 for use in WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blotting: 1:200 using ECL on rat heart membranes. See suggested protocol.
Dilutions should be made using a carrier protein such as BSA (1-3%).
Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, and 0.05% sodium azide. Reconstitute with 50 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months from date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Lysakowski et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(27), 10101-10114 (2011-07-08)
Many primary vestibular afferents form large cup-shaped postsynaptic terminals (calyces) that envelope the basolateral surfaces of type I hair cells. The calyceal terminals both respond to glutamate released from ribbon synapses in the type I cells and initiate spikes that
Thomas E Finger et al.
The Journal of comparative neurology, 525(7), 1743-1755 (2017-02-06)
The ionotropic serotonin receptor, 5-HT3 , is expressed by many developing neurons within the central nervous system. Since the olfactory epithelium continues to generate new olfactory sensory neurons (OSNs) throughout life, we investigated the possibility that 5-HT3 is expressed in
Sharon L Rowan et al.
PloS one, 7(1), e29082-e29082 (2012-01-12)
Although denervation has long been implicated in aging muscle, the degree to which it is causes the fiber atrophy seen in aging muscle is unknown. To address this question, we quantified motoneuron soma counts in the lumbar spinal cord using
Shih-Lin Chang et al.
Experimental and therapeutic medicine, 13(2), 771-779 (2017-03-30)
The impact of heart failure (HF) on sinoatrial node (SAN) channel regulation and electropharmacological responses has remained elusive. The present study aimed to investigate the effects of HF on the electrical activity of SANs with and without pharmacological interventions. Action
Christopher T Frenz et al.
Journal of neurophysiology, 112(5), 1091-1104 (2014-05-30)
Olfactory sensory neurons (OSNs) fire spontaneously as well as in response to odor; both forms of firing are physiologically important. We studied voltage-gated Na(+) channels in OSNs to assess their role in spontaneous activity. Whole cell patch-clamp recordings from OSNs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service