Skip to Content
Merck
All Photos(1)

Key Documents

AB5416P

Sigma-Aldrich

Anti-Clock Antibody

Chemicon®, from rabbit

Synonym(s):

KAT13D

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$589.00

$589.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$589.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$589.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CLOCK(9575)

Specificity

Recognizes mouse Clock. The immunogen shows no significant sequence homology with dPER or other known proteins.

SPECIES REACTIVITIES: The immunogen sequence is 92% conserved in rate and 90% in human. Reactivity with other species has not been confirmed.

Immunogen

An 18 amino acid peptide mapping downstream of the serine rich region of mouse Clock (Antoch et al. 1997; King et al. 1997).

Application

Anti-Clock Antibody detects level of Clock & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Circadian Rhythm & Sleep
Western blot: 1-10 μg/mL using ECL.

ELISA: 1:10,000-1:100,000 using 50-100ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS and 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of antibody will occasionally become entrapped in the seal of the product vial. For antibodies with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service