Skip to Content
Merck
All Photos(1)

Key Documents

AB5370P

Sigma-Aldrich

Anti-Capsaicin Receptor Antibody, CT

Chemicon®, from rabbit

Synonym(s):

Vanilloid Receptor 1, VR1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$689.00

$689.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
20 μG
$689.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$689.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TRPV1(7442)

Specificity

Capsaicin Receptor (Vanilloid Receptor 1, VR1).

Immunogen

A 21 amino acid peptide corresponding to the C-terminus of the rat Capsaicin Receptor protein.
Epitope: C-terminus

Application

Anti-Capsaicin Receptor Antibody, C-terminus is an antibody against Capsaicin Receptor for use in IH.
Immunohistochemistry: 1-2 μg/mL.

The antibody reacts well with tissue from the rat dorsal root ganglia.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neurotransmitters & Receptors

Physical form

Affinity purified immunoglobulin. Lyophilized. Contains no preservative. Reconstitute with 20 μL of sterile distilled water. Centrifuge if necessary to remove any residue.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C for up to 6 months. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Jabin Fågelskiöld et al.
Islets, 4(1), 56-63 (2012-01-26)
We have studied whether functional TRPV1 channels exist in the INS-1E cells, a cell type used as a model for β-cells, and in primary β-cells from rat and human. The effects of the TRPV1 agonists capsaicin and AM404 on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service