Skip to Content
Merck
All Photos(1)

Key Documents

AB5314

Sigma-Aldrich

Anti-Carboxypeptidase H Antibody, CT

serum, Chemicon®

Synonym(s):

CPH, Carboxypeptidase E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$632.00

$632.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CPE(1363)

Specificity

C-terminus of Carboxypepdidase H.

Immunogen

Epitope: C-terminus
Synthetic peptide corresponding to the C-terminus of Carboxypepdidase H conjugated to KLH.

Application

Anti-Carboxypeptidase H Antibody, C-terminus is an antibody against Carboxypeptidase H for use in IP, WB & IC.
Immunocytochemistry on AtT-20 cell line: 1:100-1:500.

Western blot: 1:500-1:2,000. Carboxypeptidase H precursor migrates at 53 kDa while active enzyme migrates at about 48 kDa.

Immunoprecipitation: 1:250-1:1,000

Optimal working dilutions must be determined by end user.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry

Physical form

Serum with 0.02% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Neukam et al.
Cell reports, 43(3), 113836-113836 (2024-02-29)
Endocrine cells employ regulated exocytosis of secretory granules to secrete hormones and neurotransmitters. Secretory granule exocytosis depends on spatiotemporal variables such as proximity to the plasma membrane and age, with newly generated granules being preferentially released. Despite recent advances, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service