Skip to Content
Merck
All Photos(1)

Key Documents

AB5306

Sigma-Aldrich

Anti-Beta (β)-Amyloid antibody

CHEMICON®, rabbit polyclonal

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$683.00

$683.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Anti-Amyloid Antibody, β 37-42, aβ, serum, Chemicon®

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Specificity

Amyloid-beta, 37-42.

Immunogen

Amyloid-beta 37-42 conjugated to KLH.

Application

Detect Amyloid using this Anti-Amyloid Antibody, β 37-42, aβ validated for use in ELISA, IH(P).
Immunohistochemistry: 1:100-1:400. Works on formaldehyde fixed, paraffin embedded tissue.

ELISA: 1:500-1:2000.

Optimal working dilutions must be determined by end user.

Physical form

Serum. Liquid with 0.02% azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beta-amyloid expression, release and extracellular deposition in aged rat brain slices.
Marksteiner, J; Humpel, C
Molecular Psychiatry null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service