Skip to Content
Merck
All Photos(1)

Key Documents

AB5046P

Sigma-Aldrich

Anti-NMDAR1 Splice Variant C1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$214.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Specific for the NMDAR1 C1 splice variant. AB5046P recognizes the C1 splice insert present in NR1 variants, 1a, 1b, 3a and 3b. {Gorter et al, (1997), "The Role of Alternative Splicing of the NMDAR1 Receptor Subunit in Synaptic Plasticity" Chapter 4, pg 99, in The Ionotropic Glutamate Receptors , Monaghan, D.T. and Weinhold, R.J. (eds), Humana Press, New Jersey) By Western blot the antibody recognizes a protein with molecular weight of 100 kDa.

Immunogen

Peptide corresponding to the C-terminus of the rat NMDAR1 C1 splice variant.

Application

Anti-NMDAR1 Splice Variant C1 Antibody is an antibody against NMDAR1 Splice Variant C1 for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:1,000 using ECL on rat brain lysate.

Immunohistochemistry: 1:1,000.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5 mM) so some residual salt may also be present. Reconstitute with 50 μL of PBS. Contains no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K D Holmes et al.
Journal of neurochemistry, 81(6), 1152-1165 (2002-06-18)
The N -methyl-D-aspartate receptor (NMDAR) is a multimeric transmembrane protein composed of at least two subunits. One subunit, NR1, is derived from a single gene and can be subdivided into three regions: the N-terminal extracellular domain, the transmembrane regions, and
Israel Pichardo-Casas et al.
Brain research, 1436, 20-33 (2011-12-27)
In recent years, microRNAs or miRNAs have been proposed to target neuronal mRNAs localized near the synapse, exerting a pivotal role in modulating local protein synthesis, and presumably affecting adaptive mechanisms such as synaptic plasticity. In the present study we
Fadia Ibrahim et al.
Cell reports, 3(2), 301-308 (2013-02-19)
RNA binding proteins (RBPs) have emerged as major causative agents of amyotrophic lateral sclerosis (ALS). To investigate the function of TAF15, an RBP recently implicated in ALS, we explored its target RNA repertoire in normal human brain and mouse neurons.
Pilar Sánchez-Blázquez et al.
The international journal of neuropsychopharmacology, 21(10), 938-948 (2018-06-04)
Several currently available animal models reproduce select behavioral facets of human mania as well as the abnormal glutamatergic neurotransmission and dysregulation of glycogen synthase kinase 3β that accompanies this disease. In this study, we addressed the therapeutic potential of ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service