Skip to Content
Merck
All Photos(1)

Key Documents

AB3795

Sigma-Aldrich

Anti-Cortactin Antibody, phospho-specific [Tyr466]

Chemicon®, from rabbit

Synonym(s):

Amplaxin, Oncogene EMS1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$843.00

$843.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$843.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$843.00


Estimated to ship on10 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr466)

Gene Information

human ... CTTN(2017)

General description

Cortactin is cytoskeleton protein that facilitates assembly of cortical actin. Cortactin is widely expressed in most adherent cells and is a prominent substrate of protein-tyrosine kinase Src in vivo and in vitro. The protein sequence of cortactin contains multiple amino-terminal tandem repeats of a unique 37-amino acid sequence, which associates with actin, and a Src homology 3 domain at the carboxyl terminus.

Specificity

The antibody detects the 80-85 kDa protein of cortactin.

Immunogen

Phosphorylated cortactin (Tyr466) synthetic peptide corresponding to mouse amino acid residues around tyrosine 466 (tyrosine 470 in the human sequence).

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Cortactin Antibody, phospho-specific [Tyr466] is validated for use in WB for the detection of Cortactin.

Physical form

Antibody is cross-adsorbed to nonphospho-peptide corresponding to the site of phosphorylation and preadsorbed with phosphopeptides to cortactin tyrosine 421 and 482 to remove cross-reactivity then affinity purified using phosphorylated cortactin (Tyr466) peptide. Antibody is supplied in phosphate-buffered saline (no Ca2+, no Mg2+), pH 7.3, containing 1 mg/ml BSA, and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C for up to one year from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanvir Bamra et al.
Frontiers in cellular and infection microbiology, 11, 641985-641985 (2021-05-14)
Leishmania secretes over 151 proteins during in vitro cultivation. Cellular functions of one such novel protein: mevalonate kinase is discussed here; signifying its importance in Leishmania infection. Visceral Leishmaniasis is a persistent infection, caused by Leishmania donovani in Indian subcontinent.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service