Skip to Content
Merck
All Photos(1)

Key Documents

AB3788

Sigma-Aldrich

Anti-Glu-Glu-epitope Tag Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$412.00

$412.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$412.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$412.00


Estimated to ship on14 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Glu-Glu-(EYMPME) epitope tag. {PNAS, 82(23):7952-54, 1985}

Immunogen

EE(EYMPME) peptide conjugated to KLH.

Application

Detect Glu-Glu-epitope Tag using this Anti-Glu-Glu-epitope Tag Antibody validated for use in IP, WB & IC.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western blot

Immunocytochemistry

Immunoprecipitation

Optimal working dilutions must be determined by the end user.

Physical form

Liquid in PBS, pH 7.2 with 0.09% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted for up to 6 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin H Hwang et al.
The Journal of biological chemistry, 288(44), 32064-32073 (2013-09-21)
Protein phosphatase 2A (PP2A) regulates almost all cell signaling pathways. It consists of a scaffolding A subunit to which a catalytic C subunit and one of many regulatory B subunits bind. Of the more than 80 PP2A isoforms, 10% use
Justin H Hwang et al.
Journal of virology, 88(20), 12055-12064 (2014-08-15)
Murine polyomavirus small t antigen (PyST) regulates cell cycle, cell survival, apoptosis, and differentiation and cooperates with middle T antigen (MT) to transform primary cells in vitro and in vivo. Like all polyomavirus T antigens, PyST functions largely via its
Ana Martín-Vega et al.
Science advances, 9(7), eadd7969-eadd7969 (2023-02-16)
RAS-ERK (extracellular signal-regulated kinase) pathway signals are modulated by scaffold proteins that assemble the components of different kinase tiers into a sequential phosphorylation cascade. In the prevailing model scaffold proteins function as isolated entities, where the flux of phosphorylation events

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service