Skip to Content
Merck
All Photos(1)

Key Documents

AB3582P

Sigma-Aldrich

Anti-Cyclic Nucleotide-Gated Channel 1 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

CNG1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$568.00

$568.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$568.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$568.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CNGA1(1259)

Specificity

Recognizes human Cyclic Nucleotide-Gated Channel 1 (CNG1). The immunogen shows no significant sequence homology with other CNG′s.

SPECIES REACTIVITIES: The immunogen sequence is 100% conserved in porcine, 80% in bovine, 70% in canine and 57% in rat. Reactivity with other species has not been confirmed.

Immunogen

A 20 amino acid peptide sequence near the N-terminus of human CNG1 (Dhallan et al. 1992; Distler et al. 1994; Bonigk et al. 1993).
Epitope: C-terminus

Application

Anti-Cyclic Nucleotide-Gated Channel 1 Antibody, C-terminus detects level of Cyclic Nucleotide-Gated Channel 1 & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10 g/mL using ECL.
ELISA: 0.5-1 g/mL using 1 g/mL control peptide per well.

Immunohistochemistry: not tested. It is recommended that the antibody be tried at 2-20 μg/mL on paraformaldehyde fixed tissue.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of antibody will occasionally become entrapped in the seal of the product vial. For antibodies with volumes of 200 μl or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service