Skip to Content
Merck
All Photos(1)

Key Documents

AB3534P

Sigma-Aldrich

Anti-Epithelial Sodium Channel-γ Antibody

Chemicon®, from rabbit

Synonym(s):

ENaC-gamma

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$810.00

$810.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$810.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$810.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SCNN1G(6340)

Specificity

Recognizes rat Epithelial Sodium Channel gamma (ENaC gamma). The immunogen shows no significant sequence homology with other ENaC subunits.

SPECIES REACTIVITIES: The immunogen sequence is 100% conserved in mouse, 92% in human, 80% in rabbit. Reactivity with other species has not been confirmed.

Immunogen

A 14 amino acid peptide sequence near the N-terminus of rat ENaC gamma (Linguela et al. 1993; Canessa et al.

1993; Snyder et al. 1994; Waldmann et al. 1995; Kreutz et al. 1997; Voilley et al.

1994; McDonald et al. 1994; Ludwig et al. 1998; Garty & Palmer 1997).

Application

Detect Epithelial Sodium Channel-γ using this Anti-Epithelial Sodium Channel-γ Antibody validated for use in ELISA & WB.
Western blot: 1-10 μg/mL using ECL. 83-90kDa bands; Masilamani et al 1999

ELISA: 1:10,000-1:100,000 using 1 mg/mL control peptide coated on well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS, containing 0.1% BSA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Masuda et al.
American journal of physiology. Renal physiology, 315(3), F653-F664 (2018-05-24)
The chronic intrinsic diuretic and natriuretic tone of sodium-glucose cotransporter 2 (SGLT2) inhibitors is incompletely understood because their effect on body fluid volume (BFV) has not been fully evaluated and because they often increase food and fluid intake at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service