Skip to Content
Merck
All Photos(1)

Key Documents

AB3487

Sigma-Aldrich

Anti-Transferrin Antibody

Chemicon®, from chicken

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$826.00

$826.00


Estimated to ship on01 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1225


Select a Size

Change View
100 μG
$826.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$826.00


Estimated to ship on01 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1225

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TF(7018)

Specificity

Transferrin

Immunogen

Human Transferrin, full length protein.

Application

Anti-Transferrin Antibody detects level of Transferrin & has been published & validated for use in ELISA & WB.
Research Category
Metabolism
Research Sub Category
Liver Physiology
Western blot: 1:1,000-1:2,000

ELISA, direct: 0.1 μg/mL

It is expected that the antibody will also work for immunocytochemistry and immunohistochemistry. Specific reactivity has not been confirmed.

Optimal working dilutions must be determined by the end user.

Physical form

Liquid in PBS, pH 7.2 containing 0.02 % sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service