Skip to Content
Merck
All Photos(1)

Documents

AB3150

Sigma-Aldrich

Anti-Fractin Antibody, CT

serum, Chemicon®

Synonym(s):

32 kDa Fragment of beta-Actin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

During apoptosis, beta actin is cleaved by activated caspase 3 between Asp244 and Gly245. This cleavage results in the release of an approximately 130 amino acid (15 kDa) fragment from the C-terminus of beta actin and a 32 kDa (244 amino acid) N-terminal fragment of actin, known as Fractin. The C-terminus of the Fractin fragment represents an apoptosis-specific neoepitope that is not antigenic in native full-length beta actin and therefore presents a novel and specific marker for distinguishing apoptotic from necrotic cells.

Specificity

By immunohistochemistry and Western blot, this antibody only recognizes the 32 kDa Fractin fragment but not full-length beta actin.

Immunogen

K-YELPD peptide representing the last five amino acids of the C-terminus of the 32 kDa Fractin fragment of beta-Actin. This sequence is absolutely conserved in Fractin fragments from human, mouse, rat, feline, porcine, ovine, bovine, canine, rabbit and guinea pig origin.

Application

Detect Fractin using this Anti-Fractin Antibody, C-terminus validated for use in WB, IC, IH(P).
Immunohistochemistry/Immunocytochemistry: 1:400-1:1000. 1 hr at 37°C or 4°C overnight. Note: If studying paraffin-embedded tissue, the use of antigen retrieval methods is recommended.

Western Blot: 1:1000-1:4000 for incubation overnight at 4°C. Recommended antibody dilution buffer: TBS with 3% BSA and 0.05% Tween 20, pH 7.4

Optimal working dilutions must be determined by end user.

Physical form

Liquid whole rabbit serum containing no preservatives.

Storage and Stability

Aliquot and store at -20°C for up to 12 months from date of receipt. Avoid repeated freeze-thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: Staurosporin treated neuronal cultures.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhouheng Ye et al.
Journal of neuropathology and experimental neurology, 77(3), 260-265 (2018-02-07)
Our previous study demonstrated caspase independent DNA fragmentation after very brief cerebral ischemia, the mechanism of which was unclear. In this study, we explore whether actin is cleaved following focal cerebral ischemia, and whether these structural changes of actin might
Peter L Peng et al.
Neuron, 49(5), 719-733 (2006-03-01)
ADAR2 is a nuclear enzyme essential for GluR2 pre-mRNA editing at Q/R site-607, which gates Ca2+ entry through AMPA receptor channels. Here, we show that forebrain ischemia in adult rats selectively reduces expression of ADAR2 enzyme and, hence, disrupts RNA
Yohan van de Looij et al.
Annals of clinical and translational neurology, 1(12), 955-967 (2015-01-13)
Lactoferrin (Lf) is an iron-binding glycoprotein secreted in maternal milk presenting anti-inflammatory and antioxidant properties. It shows efficient absorption into the brain from nutritional source. Brain injury frequently resulting from cerebral hypoxia-ischemia (HI) has a high incidence in premature infants
Eduardo F Sanches et al.
Frontiers in neurology, 9, 480-480 (2018-07-11)
Introduction: Prematurity, through brain injury and altered development is a major cause of neurological impairments and can result in motor, cognitive and behavioral deficits later in life. Presently, there are no well-established effective therapies for preterm brain injury and the
Vanessa Ginet et al.
BioFactors (Oxford, England), 42(3), 323-336 (2016-06-18)
Lactoferrin (Lf), component of maternal milk, has antioxidant, anti-inflammatory and antimicrobial properties. Neuroprotective effects of Lf on the immature brain have been recently shown in rodent models of intrauterine growth restriction and cerebral hypoxia/ischemia. Here we postulated that Lf could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service