Skip to Content
Merck
All Photos(1)

Key Documents

AB3120

Sigma-Aldrich

Anti-Angiopoietin-1 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

Ang-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$711.00

$711.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1522

Request a Bulk Order

Select a Size

Change View
50 μG
$711.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$711.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1522

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ANGPT1(284)

General description

The remodelling of the primitive vasculature in mammals to form the mature cardiovascular system includes a number of factors directing endothelial cell development. Angiopoietin-1 (mouse and human Ang-1; 498aa with 98% identity) is an angiogenic secreted protein that interacts with the endothelial specific Tie-2 receptor. It is primarily expressed in developing endothelial cells. During embryonic dvevelopment, Ang-1 binds and induces tyrosine phosphorylation of Tie-2. Ang-1 appears to play a crucial role in mediating matrix and mesenchyme. It mediates blood vessel maturation and stability. Ang-1 may play a critical role in heart development. Ang-1 deficient mice mimic the phenotype exhibited by animals deficient in Tie-2. A homolog of Ang-1, termed Angiopoietin-2 (mouse and human Ang-2, 496aa with 85% identity) may act as an antagonist for Ang-1 and Tie-2. Ang-1 and Ang-2 have ~60% sequence homology.

Specificity

Recognizes mouse Angiopoietin-1 (Ang-1). The immunogenic peptide sequence is 90% (18/20 aa) conserved in human Ang-1, 95% I pig and bovine, and has no significant sequence homology with Ang-2. Antibody crossreactivity in other species is not established.

Immunogen

20 amino acid peptide sequence near the amino-terminus of mouse Ang-1.
Epitope: N-terminus

Application

Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Angiopoietin-1 Antibody, N-terminus is validated for use in ELISA, WB for the detection of Angiopoietin-1.
Western blot: 1-10 ug/mL using ECL. Expected size of Ang-1 is ~55kDa.

ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide per well (Catalog Number AG635).

Immunohistochemistry: Not tested. It is recommended that the antibody be used at 2-20 ug/ml in formaldehyde fixed tissue.

Optimal working dilutions must be determined by end user.

Target description

55 kDa

Physical form

Affinity Purified immunoglobulin. Liquid in PBS with 0.1% BSA.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Fetal heart

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunil K Chauhan et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(7), 1606-1615 (2015-05-23)
In angiogenesis, circulating mononuclear cells are recruited to vascular lesions; however, the underlying mechanisms are poorly understood. Here, we characterize the functional role of protein tyrosine kinase 7 (PTK7)-expressing CD11b(+) mononuclear cells in vitro and in vivo using a mouse
Jinbo Li et al.
International journal of biological sciences, 18(5), 2091-2103 (2022-03-29)
Osteoblastic lineage cells (OBCs) are bone-building cells and essential component of hematopoietic niche, but mechanisms whereby bone-building and hematopoiesis-supportive activities of OBCs could be regulated simultaneously remain largely unknown. Here we found that B cell-specific Moloney murine leukemia virus integration
Karthik Shekhar et al.
Cell, 166(5), 1308-1323 (2016-08-28)
Patterns of gene expression can be used to characterize and classify neuronal types. It is challenging, however, to generate taxonomies that fulfill the essential criteria of being comprehensive, harmonizing with conventional classification schemes, and lacking superfluous subdivisions of genuine types.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service