Skip to Content
Merck
All Photos(1)

Key Documents

AB2959

Sigma-Aldrich

Anti-ADAM 9 Antibody, disintegrin domain

from rabbit

Synonym(s):

ADAM metallopeptidase domain 9 (meltrin gamma), a disintegrin and metalloproteinase domain 9 (meltrin gamma), Cellular disintegrin-related protein, Myeloma cell metalloproteinase, Metalloprotease/disintegrin/cysteine-rich protein 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$632.00

$632.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), primate (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

A disintegrin and metalloproteinase domain-containing protein 9 (ADAM 9) belongs to a family of transmembrane, membrane-anchored, disintegrin-containing proteins that are involved in several biological functions such as protein ectodomain shedding, cell fusion, and cell adhesion. ADAMs contain several characteristic domains, two of which, the protease and disintegrin domains, are noted for their homology to snake venom metalloprotease. The exact function of ADAM 9 is yet unknown, however it is widely distributed, is found within keratinocytes, and is ideally placed on the cell membrane, which some studies have suggested supports a role in cell migration and motility. Alternative splicing results in two isoforms; isoform 1 is found within the cell membrane and is considered a single-pass type 1 membrane protein, while isoform 2 is secreted. Mutations in ADAM 9 expression are causal to cone-rod dystrophy type 9 or CORD9 which is characterized by visible retinal pigment deposits and loss of cone photoreceptors leading and rod degeneration, ultimately resulting in severe loss of vision.

Specificity

Mouse and rat (90% sequence homology).

Immunogen

Epitope: disintegrin domain
KLH-conjugated linear peptide corresponding to the disintegrin domain of human ADAM 9.

Application

Detect ADAM 9 using this Anti-ADAM 9 Antibody, disintegrin domain validated for use in WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Quality

Evaluated by Western Blot in human chondrocyte cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected ADAM 9 in 10 µg of human chondrocyte cell lysate.

Target description

~ 90 kDa observed. 2 isoforms are produced by alternative splicing: Isoform 1 at 91 kDa and Isoform 2 at 72 kDa

Linkage

Replaces: AB19025

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl, with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human chondrocyte cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elmina Mammadova-Bach et al.
JCI insight, 1(14), e88245-e88245 (2016-10-05)
Metastatic dissemination of cancer cells, which accounts for 90% of cancer mortality, is the ultimate hallmark of malignancy. Growing evidence suggests that blood platelets have a predominant role in tumor metastasis; however, the molecular mechanisms involved remain elusive. Here, we
Narkhyun Bae et al.
Acta pharmacologica Sinica, 32(3), 288-294 (2011-03-05)
To examine the role of protein L-isoaspartyl O-methyltransferase (PIMT; EC 2.1.1.77) on the secretion of Aβ peptides. HEK293 APPsw cells were treated with PIMT siRNA or adenosine dialdehyde (AdOX), a broad-spectrum methyltransferase inhibitor. Under the conditions, the level of Aβ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service