Skip to Content
Merck
All Photos(1)

Key Documents

AB2290A4

Sigma-Aldrich

Anti-MAP2, Alexa Fluor 488 Conjugate Antibody

from rabbit, ALEXA FLUOR 488

Synonym(s):

Microtubule-associated protein 2, MAP-2, MAP2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$901.00

$901.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$901.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$901.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Map2(25595)

General description

The microtubule-associated protein 2 (MAP-2) is an abundant neuronal cytoskeletal protein that binds to tubulin, stabilizes microtubules (Herzog and Weber, 1978), and is an excellent marker for neurons. MAP-2 is essential for the development and maintenance of neuronal morphology (Matus, 1991). Three primary isoforms of MAP-2, the high molecular weight MAP2a and MAP2b, and the low molecular weight MAP2c (resulting from alternate splicing of the MAP2 gene), have been identified in neurours. (Chung et al., 1996). The low molecular weight isoform, MAP2c, is expressed in developing brain and is down-regulated during brain maturation, whereas the high molecular weight MAP2b is expressed in both developing and adult brain. The MAP2a appears only after brain maturation (Tucker, 1990). All these forms bind to microtubules through a domain near the carboxyl terminus that contains either three or four similar repeats of a 31-amino-acid motif (Lewis et al., 1988). MAP-2 together with MAP-4 and tau proteins belong to the family of thermostable proteins associated with microtubules.

Application

This Anti-MAP2, Alexa Fluor 488 Conjugate Antibody is validated for use in ICC, IHC for the detection of MAP2.

Quality

Evaluated by Immunocytochemistry in rat E18 cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected MAP2 in rat E18 cortex cells.

Target description

The unconjugated parent antibody (Cat. No. AB2290) has an observed molecular weight at ~260 kDa

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna M Grochowska et al.
EMBO reports, 18(6), 962-981 (2017-04-20)
Oligomeric amyloid-β (Aβ) 1-42 disrupts synaptic function at an early stage of Alzheimer's disease (AD). Multiple posttranslational modifications of Aβ have been identified, among which N-terminally truncated forms are the most abundant. It is not clear, however, whether modified species
Masahito Kawabori et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(8), 3384-3396 (2015-02-27)
Clearing cellular debris after brain injury represents an important mechanism in regaining tissue homeostasis and promoting functional recovery. Triggering receptor expressed on myeloid cells-2 (TREM2) is a newly identified receptor expressed on microglia and is thought to phagocytose damaged brain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service