Skip to Content
Merck
All Photos(2)

Key Documents

AB2263

Sigma-Aldrich

Anti-MEF2D Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

myocyte enhancer factor 2D, MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancer factor 2D), MEF2D/DAZAP1 fusion, myocyte-specific enhancer factor 2D, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusion prot

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rhesus macaque, rat, human, mouse, pig, horse

species reactivity (predicted by homology)

equine (based on 100% sequence homology), porcine (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MEF2D(4209)

General description

Myocyte-specific enhancer factor 2D is a transcriptional activator belonging to the MEF2 family. MEF2D has two domains of interest; a MADS-box domain, and a Mef2-type DNA-binding domain specifically binding to 5′-YTA[AT]4TAR-3′, the MEF2 element, which is observed in several gene types such as growth factor, muscle-specific, and stress-induced genes. MEF2D regulates various cellular processes including those within cardiac and skeletal muscle development, neuronal survival and differentiation, as well as cell growth, apoptosis and survival. MEF2D, along with ERK5, plays a vital role in neuronal survival by regulating the anti-apoptotic molecule, bcl-w. There are 6 known isoforms of MEF2D resulting from alternative splicing.

Immunogen

KLH-conjugated linear peptide corresponding to human MEF2D.

Application

Anti-MEF2D Antibody is an antibody against MEF2D for use in WB, IH(P).
Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected MEF2D in normal human breast skin and inflammatory skin tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blot in L6 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MEF2D on 10 µg of L6 cell lysate.

Target description

Observed at ~ 54 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
L6 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina A Sharlo et al.
International journal of molecular sciences, 24(1) (2023-01-09)
Muscle unloading leads to signaling alterations that cause muscle atrophy and weakness. The cellular energy sensor AMPK can regulate myofiber-type shift, calcium-dependent signaling and ubiquitin-proteasome system markers. We hypothesized that the prevention of p-AMPK downregulation during the first week of
Inna I Paramonova et al.
Pharmaceuticals (Basel, Switzerland), 14(11) (2021-11-28)
It is well known that reduced contractile activity of the main postural soleus muscle during long-term bedrest, immobilization, hindlimb unloading, and space flight leads to increased expression of fast isoforms and decreased expression of the slow isoform of myosin heavy
Kristina A Sharlo et al.
International journal of molecular sciences, 22(3) (2021-02-13)
It was observed that gravitational unloading during space missions and simulated microgravity in ground-based studies leads to both transformation of slow-twitch muscle fibers into fast-twitch fibers and to the elimination of support afferentation, leading to the "switching-off" of postural muscle
P J Nicholls et al.
eNeuro, 9(5) (2023-01-14)
The protease caspase-3 is a key mediator of apoptotic programmed cell death. But weak or transient caspase activity can contribute to neuronal differentiation, axonal pathfinding, and synaptic long-term depression. Despite the importance of sublethal, or nonapoptotic, caspase activity in neurodevelopment
Fernando Josa-Prado et al.
PloS one, 14(2), e0212857-e0212857 (2019-02-23)
Protein SUMOylation regulates multiple processes involved in the differentiation and maturation of cells and tissues during development. Despite this, relatively little is known about the spatial and temporal regulation of proteins that mediate SUMOylation and deSUMOylation in the CNS. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service