Skip to Content
Merck
All Photos(1)

Key Documents

AB1847

Sigma-Aldrich

Anti-C-X-C Chemokine Receptor 4 Antibody, extracellular loop

Chemicon®, from rabbit

Synonym(s):

CD184, CXCR4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$632.00

$632.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CXCR4(7852)

Specificity

Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors (Nimitrov 1997). Among them, CXCR4 (fusin, LESTR or HUMSTR) is a principle coreceptor for T-cell tropic strains of HIV-1 fusion and entry of human white blood cells (Feng et al. 1996; Berson et al. 1996). CXCR4 is also required for the infection by dual-tropic strains of HIV-1 and mediates CD-4 independent infection by HIV-2 (Doranz et al. 1996; Endres et al. 1996). The a-chemokine SDF-1 is the ligand for CXCR4 and prevents infection by T-tropic HIV-1 (Bleul et al. 1996; Oberlin et al. 1996). CXCR4 associates with the surface CD4-gp120 complex before HIV enters target cells (Lapham et al. 1996). CXCR4 messenger RNA levels correlated with HIV-1 permissiveness in diverse human cell types (Feng et al. 1996). Antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells (Feng et al. 1996; Endres et al. 1996; Brelot et al. 1997). The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV binding sites (Brelot et al. 1997; Lu et al. 1997).

Immunogen

Epitope: extracellular loop
Extracellular loop peptide of human CXCR4 (Berson et al. 1996; Leotscher et al. 1994).

Application

Anti-C-X-C Chemokine Receptor 4 Antibody, extracellular loop detects level of C-X-C Chemokine Receptor 4 & has been published & validated for use in WB.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western blot: 1:500 - 1000

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified IgG in PBS containing 0.02% sodium azide.

Storage and Stability

Can be refrigerated at 2-8°C in undiluted aliquots for up to 12 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CXCR4 attenuates cardiomyocytes mitochondrial dysfunction to resist ischaemia-reperfusion injury.
Cai, WF; Kang, K; Huang, W; Liang, JL; Feng, YL; Liu, GS; Chang, DH; Wen, ZL; Paul, C; Xu et al.
Journal of Cellular and Molecular Medicine null
Akt/hypoxia-inducible factor-1? signaling deficiency compromises skin wound healing in a type 1 diabetes mouse model.
Jing, L; Li, S; Li, Q
Experimental and Therapeutic Medicine null
Chih-Hung Guo et al.
Marine drugs, 19(4) (2021-04-04)
Fish oil (FO) and selenium (Se) possess antiangiogenic potential in malignant tumors. This study aimed to determine whether combination of FO and Se enhanced treatment efficacy of low-dose antiangiogenic agent Avastin (bevacizumab) in a dose-dependent manner and targeted multiple signaling
Yung-Ming Chang et al.
Journal of cellular physiology, 234(7), 12042-12050 (2018-12-06)
Adipose-derived mesenchymal stem cells (ADMSCs) are easily accessible and are attractive mesenchymal stem cells for use in regenerative medicine; however their application is frequently restricted due to various challenges present in the environment they are administered. Therefore ADMSCs are preferably
Heidi Schabath et al.
Journal of cell science, 119(Pt 2), 314-325 (2006-01-05)
CD24 is a small, heavily glycosylated cell-surface protein which is linked to the membrane via a glycosyl-phosphatidylinositol (GPI-) anchor and therefore localizes in lipid rafts. CD24 is widely used as a cell-lineage marker for hematopoietic cells. CD24 is also expressed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service