Skip to Content
Merck
All Photos(1)

Key Documents

AB1785P

Sigma-Aldrich

Anti-Dopamine D3 Receptor Antibody, cytoplasmic domain

Chemicon®, from rabbit

Synonym(s):

D3R

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$888.00

$888.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$888.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$888.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DRD3(1814)

Specificity

Specific for Dopamine Receptor D3. Human Dopamine D3 receptor is a 400 aa, G-protein coupled and transmembrane receptor protein (Giros et al. 1990, 1991; Schmauss et al. 1993; Liu et al. 1994; Fu et al. 1995; Sokoloff et al. 1990; Livingstone et al. 1992). The immunogen peptide shows no significant sequence similarity with other dopamine receptors (D1, D2, D4 or D5).

Immunogen

A 19 amino acid peptide sequence from the human D3 receptor within the 3rd cytoplasmic domain.
Epitope: cytoplasmic domain

Application

Detect Dopamine D3 Receptor using this Anti-Dopamine D3 Receptor Antibody, cytoplasmic domain validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1-10 μg/mL (Chemiluminescence technique) Approximately 100-400 μg of membrane protein/lane should be loaded to visualize the receptor band.

Immunohistochemistry: 1-20 μg/mL on paraformaldehyde fixed sections.

ELISA: 0.5-1.0 μg/mL (1 μg/mL immunogen peptide (Cat. Number AG224)/well)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.5 with 0.1% BSA. No preservatives

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay Chandrasekar et al.
Molecular and cellular neurosciences, 42(4), 350-362 (2009-08-26)
MicroRNAs play key regulatory roles in cellular processes including neurogenesis, synapse development and plasticity in the brain. Psychostimulants induce strong neuroadaptive changes through a surfeit of gene regulatory mechanisms leading to addiction. MicroRNA profiling for identifying miRNAs regulating cocaine-induced, plasticity-related
Frederik Ravnkilde Marlet et al.
Biochimica et biophysica acta. Molecular basis of disease, 1870(6), 167212-167212 (2024-05-16)
Parkinson's Disease (PD) is characterised by the loss of dopaminergic neurons and the deposition of protein inclusions called Lewy Bodies (LBs). LBs are heterogeneous structures composed of protein and lipid molecules and their main constituent is the presynaptic protein α-synuclein.
Jian-Da Wang et al.
Autophagy, 11(11), 2057-2073 (2015-12-10)
Autophagy dysfunction is implicated in the pathogenesis of Parkinson disease (PD). BECN1/Beclin 1 acts as a critical regulator of autophagy and other cellular processes; yet, little is known about the function and regulation of BECN1 in PD. In this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service