Skip to Content
Merck
All Photos(1)

Key Documents

AB16528

Sigma-Aldrich

Anti-NIK Antibody, CT

Chemicon®, from rabbit

Synonym(s):

NFkB inducing Kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$500.00

$500.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$500.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAP3K14(9020)

Specificity

Nuclear factor kappa B (NF-kB) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kB mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNFalpha, LPS and mitogens. A serine/threonine protein kinase which mediates NF-kB activation by IL-1, TNFa and CD95 was identified recently and designated NIK (for NF-kB inducing kinase) (Malinin et al. 1997). NIK is an activator of IkB kinase alpha and beta (IKKalpha and IKKbeta) (Regnier et al. 1997; Woronicz et al. 1997; Ling et al. 1998; Nakano et al. 1998). Therefore, NIK is a key molecule in the NF-kB signaling pathway leading to the induction of a variety of gene expression in response to proinflammatory cytokines and bacteria products.

Immunogen

Epitope: C-terminus
Rabbit anti-NIK polyclonal antibody was raised against a peptide corresponding to amino acids 931 to 947 of human NIK (Malinin et al. 1997).

Application

Anti-NIK Antibody, C-terminus is an antibody against NIK for use in WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
Western blot

293 cell lysate can be used as positive control.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service